Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Paxillin Antibodies

          Invitrogen

          Phospho-Paxillin (Tyr118) Polyclonal Antibody

          Advanced Verification
          45 Published Figures
          101 References
          View all (67) Paxillin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-Paxillin (Tyr118) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (45)
          • Advanced Verification (2)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 49

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Paxillin (Tyr118) Antibody (44-722G) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of COS-7 (Lane 1), COS-7 treated with 100 uM Pervanadate for 1 hour (Lane 2), A431 (Lane 3), A431 treated with 100 uM Pervanadate for 1 hour (Lane 4), HeLa (Lane 5) and HeLa treated with 100 uM Pervanadate for 1 hour (Lane 6).The blots were probed with Anti-Phospho-Paxillin pTyr118 Rabbit Polyclonal Antibody (Product # 44-722G, 1:500 ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Paxillin (Tyr118) Antibody in Immunohistochemistry (IHC)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody in Western Blot (WB)
          Phospho-Paxillin (Tyr118) Antibody
          Phospho-Paxillin (Tyr118) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-Paxillin (Tyr118) Polyclonal Antibody

          Product Details

          44-722G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 48 publications 48 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 42 publications 42 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Chicken, Dog, Human, Mouse, Pig, Rat, Xenopus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human paxillin that contains tyrosine 118.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533733

          Product Specific Information

          The antibody has been negatively preadsorbed using i) a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated paxillin, and ii) a generic tyrosine phosphorylated peptide to remove antibody that is reactive with phosphotyrosine, irrespective of the sequence. The final product is generated by affinity chromatography using a paxillin-derived peptide that is phosphorylated at tyrosine 118.

          Positive controls used with this antibody in western blotting were normal mouse mammary gland (NMuMG) cells, NMuMG transfected with EGFP-tagged paxillin +/- TGFbeta, and human epidermoid carcinoma cells (A431) +/- EGF.

          Target Information

          Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ16691; hCG_1778014; PAX-1; PAXI; paxillin alpha; testicular tissue protein Li 134; unnamed protein product

          View more View less

          Gene Aliases: Pax

          View more View less

          UniProt ID: (Human) P49023, (Mouse) Q8VI36

          View more View less

          Entrez Gene ID: (Human) 5829, (Mouse) 19303

          View more View less

          Function(s)
          protein binding beta-catenin binding vinculin binding protein phosphatase binding neuropilin binding MAP-kinase scaffold activity integrin binding protein kinase binding receptor signaling complex scaffold activity macromolecular complex binding BH4 domain binding
          Process(es)
          cell adhesion signal transduction signal complex assembly transforming growth factor beta receptor signaling pathway cell migration substrate adhesion-dependent cell spreading cellular response to reactive oxygen species endothelial cell migration positive regulation of stress fiber assembly growth hormone receptor signaling pathway MAPK cascade cytoskeleton organization integrin-mediated signaling pathway regulation of cell shape peptidyl-tyrosine phosphorylation lamellipodium assembly lipopolysaccharide-mediated signaling pathway positive regulation of angiogenesis positive regulation of protein kinase activity focal adhesion assembly branching morphogenesis of an epithelial tube positive regulation of substrate adhesion-dependent cell spreading response to peptide
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5d76f9f486-9kkwj:80/100.66.77.245:80.
          git-commit: 08b979eaab39376f31a39358ffa32de998e3c7f7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.6.0-offline-prod