Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control for Western blot is HUVEC cells.
The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: non-specific serine/threonine protein kinase; Oncogene PIM1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; Serine/threonine-protein kinase pim-1
Gene Aliases: PIM; Pim-1; PIM1
UniProt ID: (Human) Q5T7H7, (Rat) P26794, (Mouse) P06803
Entrez Gene ID: (Human) 5292, (Rat) 24649, (Mouse) 18712
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support