Applications | Tested Dilution | Publications |
---|---|---|
Dot blot (DB) |
1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S24 of human RGS19 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
Lot-specific |
Purification |
Protein A, Antigen affinity chromatography |
Storage buffer |
PBS |
Contains |
0.09% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2178055 |
RGS19 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G-alpha subfamily 1 members, predominantly to G(i)-alpha-3. Activity on G(z)-alpha is inhibited by phosphorylation and palmitoylation of the G-protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G alpha interacting protein; G protein signalling regulator 19; G-alpha-interacting protein; GAIP; GAIP protein; GS19; guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein; Regulator of G-protein signaling 19; regulator of G-protein signalling 19; RGS19
Gene Aliases: GAIP; GNAI3IP; RGS19; RGSGAIP
UniProt ID: (Human) P49795
Entrez Gene ID: (Human) 10287
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support