Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This purified antibody is directed against human SUMO Activating Enzyme E1 protein. This antibody is specific for human SAE1 protein phosphorylated at S185. A BLAST analysis using the sequence of the immunizing peptide was used to suggest that this antibody would react with SUMO Activating Enzyme E1 protein from human (100%), bovine, dog, chimpanzee (96%), mouse (93%), and rat (92%) based on a high degree of sequence homology. Cross reactivity against this protein from other sources has not been determined.
Posttranslational modification of proteins by the addition of the small protein SUMO (see SUMO1), or sumoylation, regulates protein structure and intracellular localization. SAE1 and UBA2. form a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins (Okuma et al., 1999).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator of SUMO1; FLJ3091; sentrin/SUMO-activating protein AOS1; SUMO-1 activating enzyme E1 N subunit; SUMO-1 activating enzyme subunit 1; SUMO-activating enzyme subunit 1; SUMO-activating enzyme subunit 1, N-terminally processed; ubiquitin-like (sentrin) activating enzyme E1A; ubiquitin-like 1 (sentrin) activating enzyme E1A; ubiquitin-like 1 (sentrin) activating enzyme subunit 1; Ubiquitin-like 1-activating enzyme E1A; ubiquitin-like protein SUMO-1 activating enzyme
Gene Aliases: 2400010M20Rik; 2610044L12Rik; AL033372; AOS1; AW743391; D7Ertd177e; HSPC140; SAE1; SUA1; Ubl1a1; UBLE1A
UniProt ID: (Human) Q9UBE0, (Bovine) A2VE14, (Mouse) Q9R1T2, (Rat) Q6AXQ0
Entrez Gene ID: (Dog) 476425, (Human) 10055, (Bovine) 505512, (Mouse) 56459, (Rat) 308384
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support