OPA1-03091 detects p-Src from human, mouse, and chicken tissues and cells.
OPA1-03091 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~60 kDa protein representing p-Src from platelet-derived growth factor (PDGF) treated NIH-3T3 cell extract. Previous lots of this antibody have been used in immunostaining.
OPA1-03091 immunizing phosphopeptide was derived from human Src that contains phosphorylated tyrosine 418.
SRC is a non-receptor kinase involved in the regulation of growth and differentiation of eukaryotic cells. SRC associates with the cytoplasmic region of various cell surface receptors to facilitate signal transduction, and is also involved in cytoskeletal organization, cell cycle regulation, and apoptosis. In humans, the gene encoding SRC is present on chromosome 20. Activity of SRC is regulated by two tyrosine residues (Tyr 416 and Tyr527), and phosphorylation of these two sites have opposing effects. Phosphorylation of Tyr416 in the activation loop of the kinase domain upregulates enzyme activity while the phosphorylation of Tyr527 decreases enzyme activity. Mutations in the SRC gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same SRC protein have been found thus far.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-src protein, C terminus; EC 2.7.10.2; Neuronal proto-oncogene tyrosine-protein kinase Src; non-tyrosine protein kinase; ORF; OTTHUMP00000174476; OTTHUMP00000174477; OTTHUMP00000174478; p60-Src; pp60c-scr; pp60c-src; pp60src; Proto-oncogene c-Src; Proto-oncogene tyrosine-protein kinase Src; protooncogene SRC, Rous sarcoma; RP5-823N20.1; src downstream region; tyrosine kinase pp60c-src; tyrosine-protein kinase SRC-1; Unknown; v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog; v-src sarcoma viral oncogene
Gene Aliases: ASV; AW259666; c-SRC; p60-Src; PP60C-SCR; pp60c-src; SDR; SRC; SRC1; THC6
UniProt ID: (Human) P12931, (Mouse) P05480
Entrez Gene ID: (Chicken) 396442, (Human) 6714, (Mouse) 20779
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support