Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunocytochemistry (ICC/IF) |
- | View 4 publications 4 publications |
ELISA (ELISA) |
- | View 1 publication 1 publication |
Immunoprecipitation (IP) |
Assay-dependent | View 1 publication 1 publication |
Miscellaneous PubMed (Misc) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Published species |
Human, Rat, Tag |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human TTK that contains threonine 33 and serine 37. |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Antigen affinity chromatography |
Storage buffer |
Dulbecco's PBS, pH 7.3, with 1mg/mL BSA, 50% glycerol |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2533594 |
This gene encodes a constant regulatory subunit of protein phosphatase 2. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. This gene encodes an alpha isoform of the constant regulatory subunit A. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 96; Dual specificity protein kinase TTK; monopolar spindle 1 kinase; monopolar spindle 1-like 1; OTTHUMP00000016778; OTTHUMP00000220743; phosphotyrosine picked threonine kinase; Phosphotyrosine picked threonine-protein kinase; PYT
Gene Aliases: CT96; ESK; MPH1; MPS1; MPS1L1; PYT; TTK
UniProt ID: (Human) P33981
Entrez Gene ID: (Human) 7272
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support