Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
MA1-750 detects prion protein (PrP) protein from a variety of species that have the conserved n-IHFG-c epitope, including agriculturally important animal species such as sheep, bovine, deer, and elk. MA1-750 has been successfully used in Western blot, immunohistochemistry and ELISA procedures. By Western blot, this antibody detects a 33-35 kDa protein from normal animals and a 27-30 kDa protein which represents PrP in brain protease treated tissue extracts from infected animals. Immunohistochemical staining of PrP(Sc) in infected sheep brain with MA1-750 results in intense staining of the spongiform lesions. In immunohistochemical procedures this antibody detects only the PrP(Sc) and not PrP(C). The MA1-750 immunogen is a synthetic peptide corresponding to residues S(146) R P L I H F G S D Y E D R(159) of bovine PrP. This peptide (Cat. # PEP-051) is available for use in neutralization and control experiments. MA1-750 is known to specifically recognize a conserved epitope of the PrP(Sc) protein comprising the amino acids n-IHFG-c.
Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alternative prion protein; AltPrP; ASCR; BSE; CD230; CD230 antigen; Major prion protein; Major scrapie-associated fibril protein 1; MGC26679; PK resistant core; prion protein; prion protein (p27-30) (Creutzfeldt-Jakob disease, Gerstmann-Strausler-Scheinker syndrome, fatal familial insomnia); prion protein precursor PrP; prion protein PrP; prion protein variant a; prion protein variant b; prion-related protein; PRNP; PrP; PrP27-30; PrP33-35C; PrPc; TSE; unnamed protein product
Gene Aliases:
AA960666; AI325101; ALTPRP; ASCR; CD230; CJD; GSS; KURU; p27-30; PRIP; prn; Prn-i; Prn-p; PRNP; PRP; PrP27-30; PrP33-35C; PrP
UniProt ID: (Human) P04156, (Bovine) P10279, (Sheep) P23907, (Mouse) P04925
Entrez Gene ID: (Human) 5621, (Bovine) 281427, (Sheep) 493887, (Mouse) 19122
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support