Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes native human PSG protein expressed in transfected mammalian cells.
Human pregnancy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregnancy thus protecting the semiallotypic fetus from rejection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD66 antigen-like family member F; CD66f; Fetal liver non-specific cross-reactive antigen 1/2; FL-NCA-1/2; FLJ90598; FLJ90654; pregnancy-specific B-1 glycoprotein; Pregnancy-specific beta-1 glycoprotein C/D; Pregnancy-specific beta-1-glycoprotein 1; PS-beta-C/D; PS-beta-G-1; PSG 1; PSG IIA-d; PSG-1d; PSG95; PSGIIA-a; PSGIIA-b; PSGIIA-c; SP 1
Gene Aliases: B1G1; PSBG1; PSG1; PSGGA
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support