Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Protocadherin Gamma Antibodies

          Rockland

          Protocadherin Gamma (pan) Monoclonal Antibody (S159-5), FITC

          View all (16) Protocadherin Gamma antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Protocadherin Gamma (pan) Monoclonal Antibody (S159-5), FITC

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Protocadherin Gamma (pan) Monoclonal Antibody (S159-5), FITC

          Product Details

          200-302-W68

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          S159-5

          Immunogen

          Anti-Protocadherin Gamma (pan) Antibody was produced by repeated immunization of mice with a fusion protein containing amino acids 808-931 of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins.
          View immunogen

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • Biotin 
          • HRP
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3' RACE product; Pan-Gamma-Protocadherin-A; PCDH gamma A; PCDH-gamma-A; PCDH-gamma-A4; PCDHGA; pcdhga4; protocadherin 13; protocadherin 2; protocadherin gamma a1; protocadherin gamma A12; protocadherin gamma a2; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma b7; protocadherin gamma C-VI; protocadherin gamma c5; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; protocadherin gamma-A1; protocadherin gamma-A10; protocadherin gamma-A11; protocadherin gamma-A12; protocadherin gamma-A2; protocadherin gamma-A3; Protocadherin gamma-A4; protocadherin gamma-A5; protocadherin gamma-A6; protocadherin gamma-A6-like; protocadherin gamma-A7; protocadherin gamma-A8; protocadherin gamma-A9; protocadherin gamma-B1; protocadherin gamma-B2; protocadherin gamma-B4; protocadherin gamma-B4-like; protocadherin gamma-B5; protocadherin gamma-B6; protocadherin gamma-B6-like; protocadherin gamma-B7; protocadherin gamma-C3; protocadherin gamma-C4; protocadherin gamma-C5

          View more View less

          Gene Aliases: mKIAA0588; mKIAA4054; pc2c; PC43; Pcdh13; Pcdh2; Pcdhga10temp; Pcdhga3; Pcdhga4; PCDHGB4; PCDHGB5; Pcdhgb7

          View more View less

          UniProt ID: (Mouse) Q91XY4

          View more View less

          Entrez Gene ID: (Rat) 102552497, (Rat) 102552705, (Rat) 102552796, (Rat) 553129, (Mouse) 93709, (Rat) 498849, (Mouse) 93722, (Rat) 252897, (Mouse) 93723, (Rat) 498850, (Mouse) 93724, (Mouse) 93710, (Rat) 498846, (Mouse) 93711, (Rat) 498847, (Mouse) 93712, (Rat) 252894, (Mouse) 93713, (Rat) 291637, (Mouse) 93714, (Mouse) 93715, (Rat) 291635, (Rat) 364843, (Mouse) 93716, (Rat) 252895, (Mouse) 93717, (Mouse) 93699, (Mouse) 93700, (Mouse) 93701, (Mouse) 93702, (Rat) 680448, (Mouse) 93703, (Mouse) 93704, (Rat) 364844, (Mouse) 93706, (Rat) 116782, (Mouse) 93707, (Mouse) 93708, (Rat) 641525

          View more View less

          Function(s)
          calcium ion binding protein binding molecular_function poly(A) RNA binding cadherin
          Process(es)
          homophilic cell adhesion via plasma membrane adhesion molecules spermatogenesis biological_process calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules endosome to lysosome transport lipid tube assembly involved in organelle fusion cell adhesion synapse organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0