Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RABEP1 gene ontology annotations related to this gene include endocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurocrescin; Rab GTPase-binding effector protein 1; RAB5 effector protein; rabaptin 5; Rabaptin-4; Rabaptin-5; Rabaptin-5alpha; Renal carcinoma antigen NY-REN-17
Gene Aliases: RAB5EP; RABEP1; RABPT5; RABPT5A
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support