Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAAPVAAD DDERRRRPGA ALEDSRSQEG ANGEAESGEL SRLRAELAGA LAEMETMKAV AEVSESTKAE AVAAVQRQCQ EEVASLQAIL KDSISSYEAQ ITALKQERQQ QQQDCEEKER ELGRLKQLLS RAYPLDSLEK QMEKAHEDSE KLREIVLPME KEIEELKAKL LRAEELIQEI QRRPRHAPSL HGSTELLPLS RDPSPPLEPL EELSGDGGPA AEAFAHNCDD SASISSFSLG GGVGSSSSLP QSRQGLSPEQ EETASLVSTG TLVPEGIYLP PPGYQLVPDT QWEQLQTEM
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fos-related antigen; MP13 protein; Rab GTPase-binding effector protein 2; Rabaptin-5 beta; Rabaptin-5beta
Gene Aliases: FRA; MP13; RABEP2; RABPT5B
UniProt ID: (Human) Q9H5N1, (Rat) Q62835
Entrez Gene ID: (Human) 79874, (Rat) 80754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support