Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with Monkey, Horse, Pi and Cat
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Rad1 is a component of a heterotrimeric PCNA-like complex that contains Rad9 and Hus1 proteins. This complex is involved in the 9-1-1 cell cycle checkpoing response complex that plays a major role in DNA repair, by associating with Rad17 and several components of the PCNA-loading heteropentamer replication factor C. Mutants has been shown to partially restore radiation resistance and G2-checkpoint activity. It has also been shown to possess exonuclease activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell cycle checkpoint protein Hrad1; Cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; RAD1 checkpoint clamp component; RAD1 homolog; Rad1-like DNA damage checkpoint protein
Gene Aliases: HRAD1; RAD1; REC1
UniProt ID: (Human) O60671
Entrez Gene ID: (Human) 5810
Molecular Function: DNA metabolism protein exodeoxyribonuclease nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support