Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: CGLCCHKHCR DQVKVECKKR PGAKGDAGPP GAPVPSTPAP HASCGSEENH SYTLSLEPET GCQLRHAWTQ TESPHPSWET DTVPCPVMDP PSTASSKLDS
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: guanyl nucleotide releasing protein 4; RAS guanyl-releasing protein 4
Gene Aliases: RASGRP4
UniProt ID: (Human) Q8TDF6
Entrez Gene ID: (Human) 115727
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support