Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 24
Description: The eB114 antibody reacts with the RASSF1A isoform of the tumor suppressor gene RASSF1. RASSF1A contains a Ras Association Domain (RA) and binds Ras in a GTP-dependent manner to effect apoptosis. The RASSF1A isoform undergoes epigenetic inactivation in a variety of human primary tumors. This has been associated with hypermethylation of a CpG island(s) in its promoter region. This antibody was generated against a synthetic peptide corresponding to the splice junction region of RASSF1A.
Applications Reported: The eB114 antibody has been reported for use in immunoprecipitation, and immunoblotting (WB).
Applications Tested: The eB114-10H1 has been tested by Western blotting and can be used at approximately 1-10 µg/mL. RASS1A is detected as a 45 kDa band in Western blotting of a HeLa cell extract. It is recommended that the reagent be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
This gene encodes a protein similar to the RAS effector proteins. Loss or altered expression of this gene has been associated with the pathogenesis of a variety of cancers, which suggests the tumor suppressor function of this gene. The inactivation of this gene was found to be correlated with the hypermethylation of its CpG-island promoter region. The encoded protein was found to interact with DNA repair protein XPA. The protein was also shown to inhibit the accumulation of cyclin D1, and thus induce cell cycle arrest. Seven alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac-specific ras association domain family 1 protein; pancreas-specific ras association domain family 1 protein; Ras association (RalGDS/AF-6) domain family member 1; Ras association domain-containing protein 1; RASSF1; tumor suppressor protein RDA32; WUGSC:H_LUCA12.5
Gene Aliases: 123F2; NORE2A; RASSF1; RASSF1A; RDA32; REH3P21
UniProt ID: (Human) Q9NS23
Entrez Gene ID: (Human) 11186
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support