Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RNQLQSVAAA CKVLIEFSLL RLENPDEACA VSQKHLILLI KGLCTGCSRL DRTEIITFTA MMKSAKLPQT VKTLSDVEDQ KELASPVSPE LRQKEVQ
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway. Recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. Together with clathrin, forms meshwork structures involved in membrane morphogenesis and cytoskeletal organization. Regulates integrin-mediated signaling. May play a role in activation of FAK in response to cell-matrix interactions. Mediates ubiquitination of ACLY, leading to its subsequent degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 600 kDa retinoblastoma protein-associated factor; E3 ubiquitin-protein ligase UBR4; N-recognin-4; p600; RBAF600; retinoblastoma-associated factor 600-like protein; Retinoblastoma-associated factor of 600 kDa; zinc finger UBR1-type protein 1
Gene Aliases: KIAA0462; KIAA1307; p600; RBAF600; UBR4; ZUBR1
UniProt ID: (Human) O75050
Entrez Gene ID: (Human) 23352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support