Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 20
Probable guanine nucleotide exchange factor. Putative effector of Ras and/or Rap. Associates with the GTP-bound form of Rap 1A and H-Ras in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDS-related protein; Ral guanine nucleotide dissociation stimulator-like 2; RalGDS-like 2; RalGDS-like factor; Ras-associated protein RAB2L
Gene Aliases: HKE1.5; KE1.5; RAB2L; RGL2
UniProt ID: (Human) O15211
Entrez Gene ID: (Human) 5863
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support