The recommended shelf life for this product is 1 year from date of receipt.
RNF219 (Ring Finger Protein 219, C13orf7) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ13449; OBI1; ORC ubiquitin ligase 1; RING finger protein 219
Gene Aliases: C13orf7; OBI1; RNF219
UniProt ID: (Human) Q5W0B1
Entrez Gene ID: (Human) 79596
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support