Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF31; FLJ23501; HOIL-1-interacting protein; HOIP; MGC19975; PAUL; Putative Ariadne-like ubiquitin ligase; RING finger protein 31; RING-type E3 ubiquitin transferase RNF31; Zinc in-between-RING-finger ubiquitin-associated domain protein
Gene Aliases: AL033293; BC031509; Flj10111; HOIP; mFLJ00217; Paul; RNF31; ZIBRA
UniProt ID: (Human) Q96EP0, (Mouse) Q924T7
Entrez Gene ID: (Human) 55072, (Mouse) 268749
Molecular Function: ubiquitin-protein ligase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support