Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: RELGRENRRL QDLATQLQEK HHRISLEYSE LQDKVTSAET KVLEMETTVE DLQWDIEKLR KREQKLNKHL AEALEQLNSG YYVSGSSSGF QGGQITLSMQ KFEMLNAELE ENQELANSRM AELEKLQAEL QGAVRTNERL KVALRSLPEE VVRETGEYRM LQAQFSLLYN ESLQVKTQLD EARGLLLATK NSHLRHIEHM E
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 95 kDa retinoblastoma protein binding protein; 95 kDa retinoblastoma-associated protein; BRE1 E3 ubiquitin ligase homolog B; BRE1-B; DKFZp686K191; E3 ubiquitin-protein ligase BRE1B; MGC13051; Rb-associated protein; RBP95; RING finger protein 40; ring finger protein 40, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1B
Gene Aliases: AI848422; BRE1B; KIAA0661; mKIAA0661; RBP95; RNF40; STARING
UniProt ID: (Human) O75150, (Mouse) Q3U319
Entrez Gene ID: (Human) 9810, (Mouse) 233900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support