Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MWKVVNLPTD LFNSVMNVGR FTEEIEWLKF LALACSALGV TITKTLKIVC EVLSCDHNGG LPRIPFSTFQ FLYTYIAEVD GEICASHVSR MLNYIEQEVI GPDGLITVND FTQNPRVWLE
The protein encoded by this gene is found in the fibrous sheath of spermatazoa, where it interacts with rhophilin, a Rho GTPase binding protein. The encoded protein also can bind an A-kinase anchoring protein (AKAP110) and a calcium-binding tyrosine phosphorylation-regulated protein (CABYR). This protein may be involved in sperm motility and has been shown to be a cancer-testis antigen in hematologic malignancies. Several transcript variants, some protein-coding and some non-protein coding, have been found for this gene. [provided by RefSeq, Dec 2015].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 91; outer dense fiber of sperm tails 6; rhophilin-associated protein 1A; ropporin, rhophilin associated protein 1; testis secretory sperm-binding protein Li 239w
Gene Aliases: CT91; ODF6; RHPNAP1; ROPN1A; ropporin
UniProt ID: (Human) Q9HAT0
Entrez Gene ID: (Human) 54763
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support