Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Involved in the cornified cell envelope formation. Multifunctional epidermal matrix protein. Reversibly binds calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: intermediate filament-associated protein; Repetin
Gene Aliases: RPTN
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support