Suitable for use as detection reagent with MA1-80661 as capture reagent in sandwich ELISA.
Mouse anti Respiratory Syncytial Virus Nucleoprotein antibody, clone RSV3132 (B023) recognizes an epitope within the 42kD RSV nucleoprotein.
Protein Aliases: human respiratory syncytial virus; RSV; RSV NP; RSV Nuclear Protein