Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence:
HCLKHGLKA KKTFLGQNKS FWGPLELVEK LVPEAAEITA SVKDLPGLKT PVGRGRAWLR LALMQKKLSE YMKALINKKE LLSEFYEPNA LMMEEEGAII AGLLVGLNVI DANFCMKGED LDSQVGVIDF SMYLKDGNSS KGTEGDGQIT AILDQKNYVE ELNRHLNATV NNLQAKVDAL EKSNTKLTEE LAVANNRIIT LQEEMERVKE ESSYILESNR KGPKQDRTAE GQALSEARKH LKEETQLRLD VEKELEMQIS MRQEMELAMK MLEKDVCEKQ DALVSLRQQL DDLRALKHEL AFKLQSSDLG VKQKSELNSR LEEKTNQMAA TIKQLEQSEK DLVKQAKTLN SAANKLIPKH H
Implicated in the formation of a single axon by developing neurons. May inhibit the formation of additional axons by inhibition of PI3K in minor neuronal processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein RUFY3; rap2 interacting protein x; Rap2-interacting protein x; RIPx; RUN and FYVE domain-containing protein 3; Singar; Singar1; singar2; Single axon-regulated protein; Single axon-regulated protein 1; single axon-related 1
Gene Aliases: 2810428M05Rik; 6330416M07Rik; AW455998; AW538594; D5Bwg0860e; KIAA0871; RIPX; Rpipx; RUFY3; SINGAR1; ZFYVE30
UniProt ID: (Human) Q7L099, (Mouse) Q9D394, (Rat) Q5FVJ0
Entrez Gene ID: (Human) 22902, (Mouse) 52822, (Rat) 360921
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support