Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:2,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
This antibody was raised against a peptide within human RUVBL1. |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.5, with 30% glycerol |
Contains |
0.035% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2793746 |
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 49 kDa TATA box-binding protein-interacting protein; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; ECP-54; INO80 complex subunit H; NMP 238; Nuclear matrix protein 238; Pontin 52; RuvB (E coli homolog)-like 1; RuvB-like 1; RuvB-like AAA ATPase 1; TAP54-alpha; TATA binding protein interacting protein 49 kDa; TIP49a; TIP60-a; TIP60-associated protein 54-alpha
Gene Aliases: ECP-54; ECP54; INO80H; NMP 238; NMP238; PONTIN; Pontin52; RUVBL1; RVB1; TIH1; TIP49; TIP49A
UniProt ID: (Human) Q9Y265
Entrez Gene ID: (Human) 8607
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support