Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RXRA Antibodies

          Proteintech

          RXRA Recombinant Rabbit Monoclonal Antibody (240878A9)

          View all (26) RXRA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RXRA Recombinant Rabbit Monoclonal Antibody (240878A9)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          RXRA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RXRA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RXRA Antibody (83796-5-RR) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 83796-5-RR (RXRA antibody) at dilution of 1:5,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RXRA Antibody in Western Blot (WB)
          RXRA Antibody in Flow Cytometry (Flow)
          RXRA Antibody in Surface plasmon resonance (SPR)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          83796-5-RR

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:10,000
          -

          Flow Cytometry (Flow)

          0.25 µg/10^6 cells in a 100 µL suspension
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Surface plasmon resonance (SPR)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240878A9

          Immunogen

          RXRA fusion protein Ag15651

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: LNGVLKVPA HPSGNMASFT KHICAICGDR SSGKHYGVYS CEGCKGFFKR TVRKDLTYTC RDNKDCLIDK RQRNRCQYCR YQKCLAMGMK REAVQEERQR GKDRNENEVE STSSANEDMP VERILEAELA VEPKTETYVE ANMGLNPSSP NDPVTNICQA ADKQLFTLVE WAKRIPHFSE LPLDDQVILL RAGWNELLIA SFSHRSIAVK DGILLATGLH VHRNSAHSAG VGAIFDRVLT ELVSKMRDMQ MDKTELGCLR AIVLFNPDSK GLSNPAEVEA LREKVYASLE AYCKHKYPEQ PGRFAKLLLR LPALRSIGLK CLEHLFFFKL IGDTPIDTFL MEMLEAPHQM T

          Target Information

          Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: nuclear receptor subfamily 2 group B member 1; ORF-3; retinoid X nuclear receptor alpha; retinoid X receptor; rxr alpha; RXR alpha 1; unnamed protein product

          View more View less

          Gene Aliases: 9530071D11Rik; NR2B1; RXR-alpha; RXRalpha; RXRalpha1

          View more View less

          UniProt ID: (Human) P19793, (Mouse) P28700

          View more View less

          Entrez Gene ID: (Human) 6256, (Mouse) 20181

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding transcription cofactor binding transcription coactivator binding retinoic acid binding DNA binding double-stranded DNA binding transcription factor activity, sequence-specific DNA binding steroid hormone receptor activity transcription cofactor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding zinc ion binding enzyme binding chromatin DNA binding peptide binding identical protein binding vitamin D receptor binding sequence-specific DNA binding retinoic acid-responsive element binding DBD domain binding LBD domain binding vitamin D response element binding sequence-specific double-stranded DNA binding protein domain specific binding retinoic acid receptor binding macromolecular complex binding thyroid hormone receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development placenta development maternal placenta development positive regulation of thyroid hormone receptor signaling pathway heart morphogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter nervous system development heart development embryo implantation negative regulation of cell proliferation hormone-mediated signaling pathway gene expression negative regulation of gene expression cell differentiation monocyte differentiation positive regulation of bone mineralization intracellular steroid hormone receptor signaling pathway regulation of myelination response to retinoic acid peroxisome proliferator activated receptor signaling pathway mRNA transcription from RNA polymerase II promoter camera-type eye development positive regulation of apoptotic process positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of translational initiation by iron retinoic acid receptor signaling pathway cell maturation response to glucocorticoid cardiac muscle cell differentiation ventricular cardiac muscle tissue morphogenesis ventricular cardiac muscle cell differentiation cardiac muscle cell proliferation mesenchyme development secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development regulation of branching involved in prostate gland morphogenesis angiogenesis involved in coronary vascular morphogenesis visceral serous pericardium development positive regulation of vitamin D receptor signaling pathway cellular response to low-density lipoprotein particle stimulus nucleus-associated proteasomal ubiquitin-dependent protein catabolic process positive regulation of lipoprotein transport positive regulation of cholesterol efflux bile acid and bile salt transport regulation of lipid metabolic process positive regulation of lipid metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-vjw2z:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline