Invitrogen
Applications | Tested Dilution |
---|---|
Immunocytochemistry (ICC/IF) |
0.1-1 µg/mL |
Product Specifications | |
---|---|
Species Reactivity |
Goat |
Host/Isotype |
Rabbit / IgG |
Class |
Recombinant Polyclonal |
Type |
Secondary Antibody |
Immunogen |
Recombinant full-length protein |
Conjugate |
Alexa Fluor™ 555
|
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Gravity column chromatography |
Storage buffer |
PBS |
Contains |
5mM sodium azide |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
RRID |
AB_2536081 |
Target |
IgG |
Antibody Form |
Whole Antibody |
The sensitivity and specificity of each lot is confirmed using ELISA.
Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig, and donkey IgG is observed.
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support