Ras is an N-ras oncogene encoding a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. The process of shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. The encoded protein, which has intrinsic GTPase activity, is activated by a guanine nucleotide-exchange factor and inactivated by a GTPase activating protein. Mutations in Ras gene have been associated with somatic rectal cancer, follicular thyroid cancer, autoimmune lymphoproliferative syndrome, Noonan syndrome, and juvenile myelomonocytic leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase NRas; N-ras protein part 4; neuroblastoma RAS viral (v-ras) oncogene homolog; Transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog
Gene Aliases: ALPS4; AV095280; CMNS; HRAS1; N-ras; NCMS; NRAS; NRAS1; NS6
UniProt ID: (Human) P01111
Entrez Gene ID: (Human) 4893, (Mouse) 18176
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support