Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MACARPLISV YSEKGESSGK NVTLPAVFKA PIRPDIVNFV HTNLRKNNRQ PYAVSELAGH QTSAESWGTG RAVARIPRVR GGGTHRSGQG AFGNMCRGGR MFAPTKTWRR WHRRVNTTQK RYAICSALAA SALPALVMSK GHRIEEVPEL PLVVEDKVEG YKKTKEAVLL LKKLKAWNDI KKVYASQRMR AGKGKMRNRR RIQRRGPCII YNEDNGIIKA FRNIPGITLL NVSKLNILKL APGGHVGRFC IWTESAFRKL DELYGTWRKA ASLKSNYNLP MHKMINTDLS RILKSPEIQR ALRAPRKKIH RRVLKKNPLK NLRIMLKLNP YAKTMRRNTI LRQARNHKLR VDKAAAAAAA LQAKSDEKAA VAGKKPVVGK KGKKAAVGVK KQKKPLVGKK AAATKKPAPE KKPAEKKPTT EEKKPAA
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L4E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L1; 60S ribosomal protein L4; Large ribosomal subunit protein uL4
Gene Aliases: L4; RPL1; RPL4
UniProt ID: (Human) P36578
Entrez Gene ID: (Human) 6124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support