Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: 293T
Immunogen sequence: MDPKISEMHP ALRLVDPQIQ LAVTRMENAV GRDQNNVGPK VYPIILRLGS PLSLNMARKT LNSLEDKAFQ LTPIAVQMTK LATTEELPDE FVVVTVK
ORF9b plays a role in the inhibition of host innate immune response by targeting the mitochondrial-associated adapter MAVS. Mechanistically, usurps the E3 ligase ITCH to trigger the degradation of MAVS, TRAF3, and TRAF6. In addition, causes mitochondrial elongation by triggering ubiquitination and proteasomal degradation of dynamin-like protein 1/DNM1L.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COVID-19 ORF9b; COVID19 ORF9b; ORF9b; SARS-CoV-2 Open Reading Frame 9b; SARS-COV2 ORF9b
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support