Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SARS-CoV-2 Spike Protein RBD Antibodies

          Invitrogen

          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Recombinant Rabbit Monoclonal Antibody (HL1003)

          View all (181) SARS-CoV-2 Spike Protein RBD antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Recombinant Rabbit Monoclonal Antibody (HL1003)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (27)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 27

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody (MA5-46917) in ICC/IF

          Immunofluorescent analysis of SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) in mock and transfected 293T cells. Cells were fixed in 4% paraformaldehyde at RT for 15 min. Green: SARS-CoV-2 (COVID-19) Spike RBD stained by SARS-CoV-2 (COVID-19) Spike RBD monoclonal antibody (Product # MA5-46917) diluted at 1:700. Blue: Fluoroshield with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Immunocytochemistry (ICC/IF)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in ELISA (ELISA)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Neutralization (Neu)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Neutralization (Neu)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Inhibition Assays (Inhibition)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Inhibition Assays (Inhibition)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Inhibition Assays (Inhibition)
          SARS-CoV-2 Spike Protein RBD (strain Wuhan-Hu-1) Antibody in Inhibition Assays (Inhibition)

          Product Details

          MA5-46917

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Neutralization (Neu)

          Assay-dependent
          -

          Inhibition Assays (Inhibition)

          13.72-10,000 ng/mL
          -
          Product Specifications

          Species Reactivity

          Virus

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          HL1003

          Immunogen

          Recombinant protein corresponding to SARS-CoV-2 (COVID-19) Spike RBD (SARS-CoV-2 (strain Wuhan-Hu-1)). The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • HRP
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2937989

          Product Specific Information

          Store as concentrated solution.

          Centrifuge briefly prior to opening vial.

          Target Information

          SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Coronavirus; COVID; Covid 19; COVID-19; COVID19; SARS; SARS Coronavirus; SARS CoV; SARS CoV-2; SARS CoV2; SARS Spike Protein; SARS-CoV-2; SARS-CoV2

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline