Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Host/Isotype |
Mouse / IgG2b |
Class |
Monoclonal |
Type |
Antibody |
Clone |
CD.E10 |
Immunogen |
Recombinant, full length human protein. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
IgG fraction |
Storage buffer |
PBS, pH 7.4 |
Contains |
0.05% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2183106 |
Recommended positive controls: 293 cells.
Predicted molecular weight is 41.5 kDa.
Stearoyl-CoA desaturase (SCD; EC 1.14.99.5) is an iron-containing enzyme that catalyzes a rate-limiting step in the synthesis of unsaturated fatty acids. The principal product of SCD is oleic acid, which is formed by desaturation of stearic acid. The ratio of stearic acid to oleic acid has been implicated in the regulation of cell growth and differentiation through effects on cell membrane fluidity and signal transduction. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5., have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA desaturase; Acyl-CoA desaturase 1; asebia; Delta(9)-desaturase; Delta(9)-desaturase 1; Delta-9 desaturase; Delta-9 desaturase 1; delta-9-desaturase; Fatty acid desaturase; Fatty acid desaturase 1; HGNC:10571; hSCD1; OTTHUMP00000020280; predicted protein of HQ0998; PRO0998; PRO1933; Stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); Stearoyl-CoA desaturase 1; stearoyl-CoA desaturase opposite strand
Gene Aliases: AA589638; ab; AI265570; FADS5; hSCD1; MSTP008; SCD; Scd-1; SCD1; SCDOS
UniProt ID: (Human) O00767, (Mouse) P13516
Entrez Gene ID: (Human) 6319, (Mouse) 20249
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support