Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SEC61B Antibodies

          Invitrogen

          SEC61B Recombinant Rabbit Monoclonal Antibody (PSH09-48)

          View all (9) SEC61B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SEC61B Recombinant Rabbit Monoclonal Antibody (PSH09-48)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (17)
          SEC61B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SEC61B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SEC61B Antibody (MA570083) in ICC/IF

          Immunocytochemistry analysis of SEC61B in C2C12 cells. Samples were incubated with SEC61B monoclonal antibody (Product # MA570083) using a dilution of 1:100 in 1% BSA in PBST overnight at 4°C followed by Goat Anti-Rabbit IgG H&L (iFluor™ 488) at a dilution of 1:1,000. Cells were fixed in 4% paraformaldehyde for 15 minutes at room temperature, permeabilized with 0.1% Triton X-100 in PBS for 15 minutes at room temperature, then blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. PBS instead of the primary antibody ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SEC61B Antibody in Immunocytochemistry (ICC/IF)
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SEC61B Antibody in Western Blot (WB)
          SEC61B Antibody in Western Blot (WB)
          SEC61B Antibody in Immunoprecipitation (IP)
          SEC61B Recombinant Rabbit Monoclonal Antibody (PSH09-48)

          Product Details

          MA570083

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Immunoprecipitation (IP)

          1-2 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH09-48

          Immunogen

          Recombinant protein within human SEC61B aa 1-96.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive control: HeLa cell lysate, HepG2 cell lysate, C2C12 cell lysate, C6 cell lysate, Bxpc-3 cell lysate, Mouse liver tissue lysate, Rat liver tissue lysate, human liver tissue, mouse liver tissue, rat liver tissue, human brain tissue, human cervical cancer tissue, mouse hippocampus tissue, mouse cerebellum tissue, mouse pancreas tissue, rat brain tissue, rat pancreas tissue.

          Subcellular location: Endoplasmic reticulum membrane.

          Recommended dilution for IF-tissue samples: 1:200

          Target Information

          The Sec61 complex is responsible for translocation of proteins across the rough endoplasmic reticulum membrane. Sec61 is composed of three subunits: alpha, beta, and gamma; that associate to form a heterotrimer. Multiple heterotrimers associate forming the ER translocon. Sec61 alpha is a polytopic protein that has ten membrane-spanning loops. Sec61 has been found to be tightly associated with membrane bound ribosomes, either directly or through adaptor proteins, and is required for assembly of membrane and secretory proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: protein translocation complex beta; protein transport protein SEC61 beta subunit; Protein transport protein Sec61 subunit beta; sec61 beta; Sec61 beta subunit; Sec61 complex, beta subunit; SEC61 translocon beta subunit; SEC61B

          View more View less

          Gene Aliases: 1190006C12Rik; AI326121; AW122942; SEC61B

          View more View less

          UniProt ID: (Human) P60468, (Mouse) Q9CQS8

          View more View less

          Entrez Gene ID: (Human) 10952, (Mouse) 66212, (Rat) 298068

          View more View less

          Function(s)
          RNA binding guanyl-nucleotide exchange factor activity protein binding ribosome binding epidermal growth factor binding poly(A) RNA binding molecular_function membrane traffic protein
          Process(es)
          SRP-dependent cotranslational protein targeting to membrane, translocation intracellular protein transport protein transport retrograde protein transport, ER to cytosol posttranslational protein targeting to membrane, translocation ERAD pathway protein import into nucleus, translocation transport ER-associated ubiquitin-dependent protein catabolic process biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline