Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:20,000-1:200,000 | View 3 publications 3 publications |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:2000 | - |
Immunocytochemistry (ICC/IF) |
1:20-1:200 | View 4 publications 4 publications |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Human, Mouse |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2F3B2 |
Immunogen |
SEPT2 Fusion Protein Ag1946 (1-361 aa encoded by BC014455) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Caprylic acid precipitation |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.1% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: MSKQQPTQF INPETPGYVG FANLPNQVHR KSVKKGFEFT LMVVGESGLG KSTLINSLFL TDLYPERVIS GAAEKIERTV QIEASTVEIE ERGVKLRLTV VDTPGYGDAI NCRDCFKTII SYIDEQFERY LHDESGLNRR HIIDNRVHCC FYFISPFGHG LKPLDVAFMK AIHNKVNIVP VIAKADTLTL KERERLKKRI LDEIEEHNIK IYHLPDAESD EDEDFKEQTR LLKASIPFSV VGSNQLIEAK GKKVRGRLYP WGVVEVENPE HNDFLKLRTM LITHMQDLQE VTQDLHYENF RSERLKRGGR KVENEDMNKD QILLEKEAEL RRMQEMIARM QAQMQMQMQG GDGDGGALGH HV (1-361 aa encoded by BC014455)
Septin 2 is a filament-forming cytoskeletal GTPase. Septin 2 is required for normal organization of the actin cytoskeleton and plays a role in the biogenesis of polarized columnar-shaped epithelium by maintaining polyglutamylated microtubules, thus facilitating efficient vesicle transport, and by impeding MAP4 binding to tubulin. It is required for the progression through mitosis, and it forms a scaffold at the midplane of the mitotic splindle required to maintain CENPE localization at kinetochores and consequently chromosome congression. During anaphase, Septin 2 may be required for chromosome segregation and spindle elongation. Septin 2 also plays a role in ciliogenesis and collective cell movements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Sep; NEDD-5; Neural precursor cell expressed developmentally down-regulated protein 5; neural precursor cell expressed, developmentally down-regulated 5; neural precursor cell expressed, developmentally down-regulated gene 5; Septin-2; Vascular endothelial cell specific protein 11
Gene Aliases: AW208991; DIFF6; hNedd5; KIAA0158; mKIAA0158; NEDD-5; NEDD5; Pnutl3; SEPT2; SEPTIN2; Vesp11
UniProt ID: (Human) Q15019, (Mouse) P42208, (Rat) Q91Y81
Entrez Gene ID: (Human) 4735, (Mouse) 18000, (Rat) 117515
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support