Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAESHLLQWL LLLLPTLCGP GTAAWTTSSL ACAQGPEFWC QSLEQALQCR ALGHCLQEVW GHVGADDLCQ ECEDIVHILN KMAKEAIFQD TMRKFLEQEC NVLPLKLLMP QCNQVLDDYF PLVIDYFQNQ TDSNGICMHL GLCKSRQPEP EQEPGMSDPL PKPLRDPLPD PLLDKLVLPV LPGALQARPG PHTQDLSEQQ FPIPLPYCWL CRALIKRIQA MIPKGALAVA VAQVCRVVPL VAGGICQCLA ERYSVILLDT LLGRMLPQLV CRLVLRCSMD DSAGPRSPTG EWLPRDSECH LCMSVTTQAG NSSEQAIPQA MLQACVGSWL DREKCKQFVE QHTPQLLTLV PRGWDAHTTC QALGVCGTMS SPLQCIHSPD L
Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventilation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18 kDa pulmonary-surfactant protein; 6 kDa protein; P SFTB3; Pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SFPB; SP-B
Gene Aliases: AI562151; PSP-B; SF-B; SFTB3; Sftp-3; SFTP3; SFTPB; SMDP1; SP-B
UniProt ID: (Human) P07988, (Mouse) P50405
Entrez Gene ID: (Human) 6439, (Mouse) 20388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support