Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Predicted to react with Mouse and Rat samples.
PRAGMIN is also known as SGK223 (sugen kinase 223). The rat homolog was originally identified in a yeast two-hybrid screen for Rnd2 effector proteins expressed in neural cells. Rat Pragmin was found to stimulate RhoA activity for the regulation of neurite outgrowth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive tyrosine-protein kinase PRAG1; Notch activation complex kinase; PEAK1-related kinase-activating pseudokinase 1; Pragma of Rnd2; Pragmin; SgK223; Sugen kinase 223; Tyrosine-protein kinase SgK223
Gene Aliases: 9830148H23; D8Ertd82e; mFLJ00269; NACK; PRAG1; PRAGMIN; RGD1311793; SGK223
UniProt ID: (Mouse) Q571I4, (Rat) D3ZMK9, (Human) Q86YV5
Entrez Gene ID: (Mouse) 244418, (Rat) 306506, (Human) 157285
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support