Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
Assay-dependent | View 1 publication 1 publication |
Immunocytochemistry (ICC/IF) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Not Applicable |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
67.8 |
Immunogen |
Recombinant protein derived from the N-terminal region of mouse Shh protein. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.4 |
Contains |
0.1% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2532212 |
Sonic Hedgehog (SHH), which is expressed only during embryogenesis, is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the Sonic Hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved, the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the developing embryo. Defects in this protein or in its signalling pathway are a cause of holoprosencephaly (HPE), a disorder in which the developing forebrain fails to correctly separate into right and left hemispheres. HPE is manifested by facial deformities. In addition, it is thought that mutations in this gene or in its signalling pathway may be responsible for VACTERL syndrome, which is characterized by vertebral defects, anal atresia, tracheoesophageal fistula with esophageal atresia, radial and renal dysplasia, cardiac anomalies, and limb abnormalities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hemimelic extra toes; HHG-1; SHH; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; ShhN; ShhNC; ShhNp; short digits; sonic hedgehog homolog; Sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein N-product
Gene Aliases: 9530036O11Rik; Dsh; HHG1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; SHH; SMMCI; TPT; TPTPS; Vhh-1
UniProt ID: (Human) Q15465, (Mouse) Q62226, (Rat) Q63673
Entrez Gene ID: (Human) 6469, (Mouse) 20423, (Rat) 29499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support