Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MCSCSLTPVP EPVLREGAEG LHCSEHLLLE KAPSYGSEGP AQRVLEGTLL EFTKDMDQLL QLTRKLESSQ PRQGEGNRTP ESQKRKSKKA TKQTLQDSFL LDLKSPPSFP VEISDRLPAA SWEGQESCWN KQTSKSEGTQ PEGTYGEQLV PELCNQSESS GEDFFLKSRL QEQDVWRRST SFYTHM
SLC12A8 is a cation/chloride cotransporter that may play a role in the control of keratinocyte proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cation-chloride cotransporter 9; solute carrier family 12 (sodium/potassium/chloride transporters), member 8; Solute carrier family 12 member 8
Gene Aliases: CCC9; SLC12A8
UniProt ID: (Human) A0AV02
Entrez Gene ID: (Human) 84561
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support