Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MASVLSYESL VHAVAGAVGS VTAMTVFFPL DTARLRLQVD EKRKSKTTHM VLLEIIKEEG LLAPYRGWFP VISSLCCSNF VYFYTFNSLK ALWVKGQHST TGKDLVVGFV AGVVNVLLTT PLWVVNTRLK LQGAKFRNED IVPTNYKGII DAFHQIIRDE GISALWNGTF PSLLLVFNPA IQFMFYEGLK RQLLKKRMKL SSLDVFIIGA VAKAIATTVT YPLQTVQSIL RFGRHRLNPE NRTLGSLRNI LYLLHQRVRR FGIMGLYKGL EAKLLQTVLT AALMFLVYEK LTAATFTVMG LKRAHQH
This protein encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SLC25A17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; unnamed protein product
Gene Aliases: PMP34
UniProt ID: (Human) O43808
Entrez Gene ID: (Human) 10478
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support