Reconstitute antibody with 50 µL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Peptide sequence is: (C)ESVSEPR KSFMYSR
Homology: Rat, human - 13/14 amino acid residues identical.
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum membrane protein; Endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV-1 protein, estrogen regulated; slc39a6 {ECO:0000312|EMBL:AAH97493.1}; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; Solute carrier family 39 member 6; Zinc transporter ZIP6; ZIP-6; Zrt- and Irt-like protein 6
Gene Aliases: Ermelin; LIV-1; LIV1; SLC39A6; ZIP6
UniProt ID: (Human) Q13433, (Mouse) Q8C145, (Rat) Q4V887
Entrez Gene ID: (Human) 25800, (Mouse) 106957, (Rat) 291733
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support