Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
In direct ELISAs, no cross-reactivity with recombinant mouse SLURP1 is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anti-neoplastic urinary protein; ANUP; ARS component B; ARS(component B)-81/S; lymphocyte antigen 6-like secreted; Secreted Ly-6/uPAR-related protein 1; secreted Ly6/uPAR related protein 1; SLURP-1
Gene Aliases: ANUP; ARS; ArsB; LY6LS; MDM; SLURP1
UniProt ID: (Human) P55000
Entrez Gene ID: (Human) 57152
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support