Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Regulator required for centriole duplication, for proper bipolar spindle formation and chromosome congression in mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 52; Coiled-coil domain-containing protein 52; spindle and centriole protein; Spindle and centriole-associated protein; Spindle and centriole-associated protein 1
Gene Aliases: CCDC52; SPICE; SPICE1
UniProt ID: (Human) Q8N0Z3
Entrez Gene ID: (Human) 152185
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support