Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
SRBD1 gene ontology annotations related to this gene include nucleobase-containing compound metabolic process; translation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H_NH0576F01.1; S1 RNA-binding domain-containing protein 1; WUGSC:H_NH0576F01.1
Gene Aliases: SRBD1
UniProt ID: (Human) Q8N5C6
Entrez Gene ID: (Human) 55133
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support