Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEQPPAPKS KLKKLSEDSL TKQPEEVFDV LEKLGEGSYG SVFKAIHKES GQVVAIKQVP VESDLQEIIK EISIMQQCDS PYVVKYYGSY FKNTDLWIVM EYCGAGSVSD IIRLRNKTLI EDEIATILKS TLKGLEYLHF MRKIHRDIKA GNILLNTEGH AKLADFGVAG QLTDTMAKRN TVIGTPFWMA PEVIQEIGYN CVADIWSLGI TSIEMAEGKP PYADIHPMRA IFMIPTNPPP TFRKPELWSD DFTDFVKKCL VKNPEQRATA TQLLQHPFIK NAKPVSILRD LITEAMEIKA KRHEEQQ (1-306 aa encoded by BC010640)
STK3 (MST2) is a STE kinase family member that acts upstream of the MAP kinase signaling pathways, and is known to be activated by the proapoptotic stimuli staurosporine and FAS ligand. STK3 is a stress-activated, pro-apoptotic kinase which, following caspase-cleavage, enters the nucleus and induces chromatin condensation followed by internucleosomal DNA fragmentation. It is a key component of the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. STK3/MST2 and STK4/MST1 are required to repress proliferation of mature hepatocytes, to prevent activation of facultative adult liver stem cells (oval cells), and to inhibit tumor formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KB-1458E12.1; Mammalian STE20-like protein kinase 2; MST-2; MST2/C; MST2/N; protein kinase homolog; serine/threonine kinase 3 (STE20 homolog, yeast); serine/threonine kinase 3 (Ste20, yeast homolog); Serine/threonine-protein kinase 3; Serine/threonine-protein kinase 3 20kDa subunit; Serine/threonine-protein kinase 3 36kDa subunit; Serine/threonine-protein kinase Krs-1; STE20-like kinase MST2
Gene Aliases: 0610042I06Rik; KRS1; Mess1; MST; MST2; Mst3; Ste20; STK3
UniProt ID: (Human) Q13188, (Rat) O54748, (Mouse) Q9JI10
Entrez Gene ID: (Human) 6788, (Rat) 65189, (Mouse) 56274
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support