Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGDIFLCKKV ESPKKNLRES KQREEDDEDP DLIYVGVEHV HRDAEVLFVG MISNSKPVVS NILNRVTPGS NSRRKKGHFR QYPAHVSQPA NHVTSMAKAI MPVSLSEGRS TDSPVTMKSS SEPGYKMSSP QVVSPSSSDS LPPGTQCLVG AMVSGGGRNE SSPDSKRLST SDINSRDSKR VKLRDGIPGV PSLAVVPSDM SSTISTNTPS QGICNSSNHV QNGVTFPWPD ANGKAHFNLT DPERASESAL AMTDISSLAS QNKTFDPKKE NPIVLLSDFY YGQHKGDGQP EQKTHTTFKC LSCVKVLKNI KFMNHMKHHL EFEKQRNDSW EDHTTCQHCH RQFPTPFQLQ CHIDSVHIAM GPSAVCKICE LSFETDQVLL QHMKDHHKPG EMPYVCQVCH YRSSVFADVE THFRTCHENT KNLLCLFCLK LFKTAIPYMN HCWRHSRRRV LQCSKCRLQF LTLKEEIEHK TKDHQTFKKP EQLQGLPSET KVIIQTSVQP GSSGMASVIV SNTDPQSSPV KTKKKTAMNT RDSRLPCSKD SS
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. SUHW1 (Suppressor of hairy wing homolog 1), also known as ZNF280A (Zinc finger protein 280A), 3'OY11.1, ZNF636 or ZNF280, is a 542 amino acid protein that contains four C2H2-type zinc fingers. Localized to the nucleus, SUHW1 is thought to function as a transcription factor that may mediate transcriptional regulation events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'OY11.1; Suppressor of hairy wing homolog 1; zinc finger protein 280; Zinc finger protein 280A; Zinc finger protein 636
Gene Aliases: 3'OY11.1; SUHW1; ZNF280; ZNF280A; ZNF636
UniProt ID: (Human) P59817
Entrez Gene ID: (Human) 129025
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support