Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SUMO1 Antibodies

          Invitrogen

          SUMO1 Recombinant Rabbit Monoclonal Antibody (5F3)

          View all (43) SUMO1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SUMO1 Recombinant Rabbit Monoclonal Antibody (5F3)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SUMO1 Antibody (MA572440) in ICC/IF

          Immunocytochemistry analysis of SUMO1 in HeLa cells. Permeabilized with Trinton X-100 at room temp for 20 min. Samples were incubated with SUMO1 Recombinant Monoclonal (Product # MA572440 using a dilution at 1:100, for 90 min at 37°C. Followed by conjugated Goat Anti-Rabbit IgG antibody (green) at 37°C for 90 min, DAPI was used to stain the cell nuclei. PBS instead of the primary antibody was used as the blank control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          SUMO1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA572440

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -

          Immunoprecipitation (IP)

          1:20-1:50
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5F3

          Immunogen

          KLH conjugated synthetic peptide derived from human Sumo 1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          40 µg

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          SUMO1 is an ubiquitin-like protein that can be covalently attached to proteins as a monomer or a lysine-linked polymer. Covalent attachment, via an isopeptide bond, to its substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I, and can be promoted by E3 ligases such as PIAS1-4, RANBP2 or CBX4. This post-translational modification on lysine residues of proteins plays a crucial role in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis and signal transduction. SUMO1 is involved, for instance, in targeting RANGAP1 to the nuclear pore complex protein RANBP2. Polymeric SUMO1 chains are also susceptible to polyubiquitination which functions as a signal for proteasomal degradation of modified proteins. SUMO1 may also regulate a network of genes involved in palate development. Mutations in the gene can result in non-syndromic orofacial cleft 10.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CG4494-PA; GAP modifying protein 1; LD07775p; Mutations: 288:OK; OTTHUMP00000202403; OTTHUMP00000202404; OTTHUMP00000202405; OTTHUMP00000202406; OTTHUMP00000202409; OTTHUMP00000202410; sentrin; Sentrin-1; similar to ubiquitin and to yeast Smt3p (suppressor of MIF2); Method: conceptual translation supplied by author; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SUMO-1; SUMO-1 related peptidase; ubiquitin-homology domain protein PIC1; ubiquitin-like 1; ubiquitin-like 1 (sentrin); ubiquitin-like protein; ubiquitin-like protein SMT3C; ubiquitin-like protein UBL1; UBL1; unnamed protein product

          View more View less

          Gene Aliases: DAP1; GMP1; OFC10; PIC1; SENTRIN; SMT3; SMT3C; SMT3H3; SMTP3; SUMO-1; UBL1

          View more View less

          UniProt ID: (Human) P63165, (Rat) Q5I0H3, (Mouse) P63166

          View more View less

          Entrez Gene ID: (Human) 7341, (Rat) 301442, (Mouse) 22218

          View more View less

          Function(s)
          RNA binding protein binding potassium channel regulator activity enzyme binding protein tag ubiquitin protein ligase binding small protein activating enzyme binding ubiquitin-like protein ligase binding transporter activator activity ubiquitin-specific protease binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription, DNA-templated protein sumoylation PML body organization positive regulation of protein complex assembly regulation of protein stability positive regulation of proteasomal ubiquitin-dependent protein catabolic process cellular response to heat negative regulation of protein import into nucleus negative regulation of action potential negative regulation of transcription, DNA-templated protein stabilization palate development negative regulation of transcription initiation from RNA polymerase II promoter cellular response to cadmium ion cellular response to hypoxia regulation of cardiac muscle cell contraction protein localization to nuclear pore regulation of calcium ion transmembrane transport DNA repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-r8ntx:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline