Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SUR1/SUR2B Antibodies

          Antibodies Inc

          SUR1/SUR2B Monoclonal Antibody (N323A/31), FL490

          View all (5) SUR1/SUR2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SUR1/SUR2B Monoclonal Antibody (N323A/31), FL490

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          SUR1/SUR2B Monoclonal Antibody (N323A/31), FL490

          Product Details

          75-298-FL490

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N323A/31

          Immunogen

          Fusion protein amino acids 1503-1545 of rat SUR2B (accession number Q63563-2) produced recombinantly in E. Coli.
          3D Epitope / Immunogen

          Conjugate

          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Excitation/Emission Max

          491/515 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Sequence of this protein is as follows: VHTILTADLV IVMKRGNILE YDTPESLLAQ EDGVFASFVR ADM

          Target Information

          Sulfonylurea receptors (SUR) are membrane proteins which are the molecular targets of the sulfonylurea class of anti-diabetic drugs whose mechanism of action is to promote insulin release from pancreatic beta cells. More specifically, SUR proteins are subunits of the inward-rectifier potassium ion channels Kir6.x (6.1 and 6.2). The association of four Kir6.x and four SUR subunits form an ion conducting channel commonly referred to as the KATP channel. The primary function of the sulfonylurea receptor is to sense intracellular levels of the nucleotides ATP and ADP and in response facilitate the open or closing its associated Kir6.x potassium channel. Hence the KATP channel monitors the energy balance within the cell.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ABC36; ABC37; ATFB12; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette transporter sub-family C member 9; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 9; ATP-binding cassette, sub-family C, member 9-like; ATP-binding cassette, subfamily C (CFTR/MRP), member 8; ATP-binding cassette, subfamily C (CFTR/MRP), member 9; CANTU; CMD1O; FLJ36852; HHF1; HI; HRINS; K-ATP channel subunit; MRP8; PHHI; sulfonylurea receptor; sulfonylurea receptor 1; Sulfonylurea receptor 2; sulfonylurea receptor 2 protein; sulfonylurea-binding protein 2; sulphonylurea receptor 1; SUR1delta2; TNDM2; unnamed protein product

          View more View less

          Gene Aliases: D930031B21Rik; Sur; SUR1; Sur2; SUR2A; SUR2B

          View more View less

          UniProt ID: (Rat) Q09429, (Rat) Q63563, (Mouse) P70170

          View more View less

          Entrez Gene ID: (Mouse) 20927, (Rat) 25559, (Rat) 25560, (Mouse) 20928

          View more View less

          Function(s)
          potassium channel activity ATP binding zinc ion binding sulfonylurea receptor activity ATPase activity cation-transporting ATPase activity ADP binding ion channel binding ABC-type transporter activity inward rectifier potassium channel activity protein binding potassium channel regulator activity transmembrane transporter activity ATPase activity, coupled to transmembrane movement of substances macromolecular complex binding potassium channel activator activity heterocyclic compound binding identical protein binding
          Process(es)
          MAPK cascade action potential blood vessel development response to hypoxia heart morphogenesis vascular process in circulatory system regulation of transcription from RNA polymerase II promoter potassium ion transport substrate-dependent cell migration, cell contraction response to stress mitochondrion organization heart development skeletal muscle tissue development blood circulation regulation of blood pressure response to xenobiotic stimulus gene expression response to activity fatty acid oxidation response to ATP response to potassium ion cellular response to potassium ion response to decreased oxygen levels vasodilation regulation of membrane potential response to hydrogen peroxide negative regulation of apoptotic process response to estrogen cellular respiration negative regulation of blood pressure ATP metabolic process fibroblast proliferation defense response to virus transmembrane transport coronary vasculature development cardiac conduction cellular response to chemical stress response to oxygen levels cellular response to calcium ion cellular response to ATP cellular response to xenobiotic stimulus potassium ion transmembrane transport circulatory system development oxygen metabolic process cardiac muscle cell contraction regulation of blood vessel diameter cation transmembrane transport regulation of potassium ion transmembrane transport response to peptide reactive oxygen species biosynthetic process response to hydrogen sulfide potassium ion import across plasma membrane cellular glucose homeostasis female pregnancy memory visual learning response to pH response to zinc ion negative regulation of low-density lipoprotein particle clearance negative regulation of angiogenesis cellular response to nutrient levels response to lipopolysaccharide positive regulation of tumor necrosis factor production response to insulin positive regulation of insulin secretion involved in cellular response to glucose stimulus positive regulation of potassium ion transport negative regulation of insulin secretion negative regulation of neurogenesis regulation of insulin secretion neuromuscular process negative regulation of glial cell proliferation glutamate secretion, neurotransmission negative regulation of neuroblast migration positive regulation of uterine smooth muscle relaxation positive regulation of voltage-gated potassium channel activity positive regulation of occluding junction disassembly negative regulation of blood-brain barrier permeability
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-br959:80/100.66.76.64:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0