Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD169; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; Sialoadhesin; Siglec-1
Gene Aliases: SIGLEC1; SN
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support