Product References
Exogenous induction of unphosphorylated PTEN reduces TGFβ-induced extracellular matrix expressions in lung fibroblasts.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
Kimura M,Hashimoto N,Kusunose M,Aoyama D,Sakamoto K,Miyazaki S,Ando A,Omote N,Imaizumi K,Kawabe T,Hasegawa Y
MA137028 was used in western blot to test if exogenous transfer of unphosphorylated phosphatase and tensin homologue deleted from chromosome 10 leads to reduce transforming growth factor beta-induced extracellular matrix expression in both epithelial cells and fibroblasts
Sun Jan 01 00:00:00 EST 2017
Altered Dermal Fibroblasts in Systemic Sclerosis Display Podoplanin and CD90.
The American journal of pathology
Nazari B,Rice LM,Stifano G,Barron AM,Wang YM,Korndorf T,Lee J,Bhawan J,Lafyatis R,Browning JL
MA137028 was used in immunohistochemistry - paraffin section to characterize alterations in fibroblast populations in the skin of patients with systemic sclerosis
Sat Oct 01 00:00:00 EDT 2016
Ductular reactions in the liver regeneration process with local inflammation after physical partial hepatectomy.
Laboratory investigation; a journal of technical methods and pathology
Suzuki Y,Katagiri H,Wang T,Kakisaka K,Kume K,Nishizuka SS,Takikawa Y
MA137028 was used in immunohistochemistry - paraffin section to examine the liver regeneration process after physical partial hepatectomy
Tue Nov 01 00:00:00 EDT 2016
Histatin-1 Expression in Human Lacrimal Epithelium.
PloS one
Shah D,Ali M,Pasha Z,Jaboori AJ,Jassim SH,Jain S,Aakalu VK
MA1-37028 was used in immunohistochemistry - paraffin section to study human lacrimal epithelium and histatin-1 expression
Tue Jul 12 00:00:00 EDT 2016
A novel vascular pattern promotes metastasis of hepatocellular carcinoma in an epithelial-mesenchymal transition-independent manner.
Hepatology (Baltimore, Md.)
Fang JH,Zhou HC,Zhang C,Shang LR,Zhang L,Xu J,Zheng L,Yuan Y,Guo RP,Jia WH,Yun JP,Chen MS,Zhang Y,Zhuang SM
MA1-37028 was used in immunohistochemistry (paraffin) to identify and characterize vessels that encapsulated tumor clusters in hepatocellular carcinoma sections.
Sat Aug 01 00:00:00 EDT 2015
The immunohistochemical profile of granular cell (Abrikossoff) tumor suggests an endomesenchymal origin.
Archives of dermatological research
Gurzu S,Ciortea D,Tamasi A,Golea M,Bodi A,Sahlean DI,Kovecsi A,Jung I
MA1-37028 was used in immunohistochemistry to identify tissue origin of the granular cell tumor using immunohistochemistry
Sun Mar 01 00:00:00 EST 2015
Myofibrotic malformation vessels: unique angiodysplasia toward the progression of hemorrhoidal disease.
Drug design, development and therapy
Li SL,Jing FY,Ma LL,Guo LL,Na F,An SL,Ye Y,Yang JM,Bao M,Kang D,Sun XL,Deng YJ
MA1-37028 was used in immunohistochemistry - paraffin section to study angiodysplasia and other pathological changes associated with symptomatic hemorrhoids
Fri Jun 03 00:00:00 EDT 2016
Primary sclerosing epithelioid fibrosarcoma of kidney with variant histomorphologic features: report of 2 cases and review of the literature.
Diagnostic pathology
Ertoy Baydar D,Kosemehmetoglu K,Aydin O,Bridge JA,Buyukeren B,Aki FT
MA1-37028 was used in immunohistochemistry to present two cases of primary sclerosing epithelioid fibrosarcoma of the kidney
Fri Oct 09 00:00:00 EDT 2015
Human pathological basis of blood vessels and stromal tissue for nanotechnology.
Advanced drug delivery reviews
Nishihara H
MA1-37028 was used in immunocytochemistry to review the pathophysiology of nanotechnology
Tue Jul 01 00:00:00 EDT 2014
Tight skin 2 mice exhibit a novel time line of events leading to increased extracellular matrix deposition and dermal fibrosis.
Matrix biology : journal of the International Society for Matrix Biology
Long KB,Artlett CM,Blankenhorn EP
MA1-37028 was used in immunohistochemistry to study the time-course of extracellular matrix deposition and dermal fibrogenesis in the murine Tsk2 model of systemic sclerosis
Mon Sep 01 00:00:00 EDT 2014
Long-term B cell depletion in murine lupus eliminates autoantibody-secreting cells and is associated with alterations in the kidney plasma cell niche.
Journal of immunology (Baltimore, Md. : 1950)
Wang W,Rangel-Moreno J,Owen T,Barnard J,Nevarez S,Ichikawa HT,Anolik JH
MA1-37028 was used in immunohistochemistry to study the effects on auto-antibody secreting cells and the kidney plasma cell niche of long-term rituximab-mediated B cell depletion in a murine lupus model
Tue Apr 01 00:00:00 EDT 2014
Human adipose tissue possesses a unique population of pluripotent stem cells with nontumorigenic and low telomerase activities: potential implications in regenerative medicine.
Stem cells and development
Ogura F,Wakao S,Kuroda Y,Tsuchiyama K,Bagheri M,Heneidi S,Chazenbalk G,Aiba S,Dezawa M
MA1-37028 was used in immunohistochemistry to study the implications for tissue engineering and regenerative medicine of the identification of a population of human adipose tissue stem cells with low telomerase that lack teratogenic activity
Tue Apr 01 00:00:00 EDT 2014
The time-dependent expression of α7nAChR during skeletal muscle wound healing in rats.
International journal of legal medicine
Fan YY,Zhang ST,Yu LS,Ye GH,Lin KZ,Wu SZ,Dong MW,Han JG,Feng XP,Li XB
MA1-37028 was used in immunohistochemistry to study the time-course and cellular localization of alpha7nAChR expression in rat skeletal muscle following contusion injury
Mon Sep 01 00:00:00 EDT 2014
Use of platelet-rich plasma solution applied with composite chondrocutaneous graft technique: an experimental study in rabbit model.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons
Sevim KZ,Yazar M,Irmak F,Tekkeşin MS,Yildiz K,Sirvan SS
MA1-37028 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of using platelet-rich plasma in composite chondrocutaneous graft procedures
Tue Jul 01 00:00:00 EDT 2014
Dynamic intersection of the longitudinal muscle and external anal sphincter in the layered structure of the anal canal posterior wall.
Surgical and radiologic anatomy : SRA
Muro S,Yamaguchi K,Nakajima Y,Watanabe K,Harada M,Nimura A,Akita K
MA1-37028 was used in immunohistochemistry to study the detailed structure of the posterior anal canal
Fri Aug 01 00:00:00 EDT 2014
A differential role for CD248 (Endosialin) in PDGF-mediated skeletal muscle angiogenesis.
PloS one
Naylor AJ,McGettrick HM,Maynard WD,May P,Barone F,Croft AP,Egginton S,Buckley CD
MA1-37028 was used in immunohistochemistry - frozen section to characterize PDGF-mediated skeletal muscle angiogenesis and the role of CD248 (Endosialin)
Tue Jun 16 00:00:00 EDT 2015
Comparative characterization of stroma cells and ductal epithelium in chronic pancreatitis and pancreatic ductal adenocarcinoma.
PloS one
Helm O,Mennrich R,Petrick D,Goebel L,Freitag-Wolf S,Röder C,Kalthoff H,Röcken C,Sipos B,Kabelitz D,Schäfer H,Oberg HH,Wesch D,Sebens S
MA1-37028 was used in immunohistochemistry - paraffin section to characterize stroma cells and ductal epithelium comparing chronic pancreatitis and pancreatic ductal adenocarcinoma
Fri Jun 12 00:00:00 EDT 2015
Transgenic mice overexpressing neuregulin-1 model neurofibroma-malignant peripheral nerve sheath tumor progression and implicate specific chromosomal copy number variations in tumorigenesis.
The American journal of pathology
Kazmi SJ,Byer SJ,Eckert JM,Turk AN,Huijbregts RP,Brossier NM,Grizzle WE,Mikhail FM,Roth KA,Carroll SL
MA1-37028 was used in immunohistochemistry to develop a neuregulin-1 overexpressing transgenic mouse model of neurofibroma-malignant peripheral nerve sheath tumor progression
Fri Mar 01 00:00:00 EST 2013
Impaired cornea wound healing in a tenascin C-deficient mouse model.
Laboratory investigation; a journal of technical methods and pathology
Sumioka T,Kitano A,Flanders KC,Okada Y,Yamanaka O,Fujita N,Iwanishi H,Kao WW,Saika S
MA1-37028 was used in immunohistochemistry to study the stromal healing of injured corneas in mice ablated for tenascin C
Fri Feb 01 00:00:00 EST 2013
Thy-1 knockdown retards wound repair in mouse skin.
Journal of dermatological science
Lee MJ,Shin JO,Jung HS
MA1-37028 was used in immunohistochemistry to study the role of Thy-1 in the repair of murine skin wounds using siRNA-mediated knockdown
Fri Feb 01 00:00:00 EST 2013
The effect of platelet-rich plasma on flap survival in random extension of an axial pattern flap in rabbits.
Plastic and reconstructive surgery
Kim HY,Park JH,Han YS,Kim H
MA1-37028 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of autologous platelet-rich plasma on axial pattern flap procedures in a rabbit model
Mon Jul 01 00:00:00 EDT 2013
Cyclic adenosine monophosphate response-element binding protein activation by mitogen-activated protein kinase-activated protein kinase 3 and four-and-a-half LIM domains 5 plays a key role for vein graft intimal hyperplasia.
Journal of vascular surgery
Nakanishi K,Saito Y,Azuma N,Sasajima T
MA1-37028 was used in immunohistochemistry to study the intimal hyperplasia of vein grafts and the roles played by CREB, MAPKAPK3 and FHL5
Tue Jan 01 00:00:00 EST 2013
Cannabinoid receptor 1 suppresses transient receptor potential vanilloid 1-induced inflammatory responses to corneal injury.
Cellular signalling
Yang Y,Yang H,Wang Z,Varadaraj K,Kumari SS,Mergler S,Okada Y,Saika S,Kingsley PJ,Marnett LJ,Reinach PS
MA1-37028 was used in immunohistochemistry to study the mechanism by which injury-induced activativation of CB1 receptors leads to downregulation of corneal TRPV1-induced inflammation
Fri Feb 01 00:00:00 EST 2013
Malignant transformation of adenomyoepithelioma of the breast by a monophasic population: a report of two cases and review of literature.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Marian C,Boila A,Soanca D,Malau M,Podeanu DM,Resetkova E,Stolnicu S
MA1-37028 was used in immunohistochemistry to report on two cases of malignant transformation of adenomyoepithelioma of the breast by a monophasic population
Mon Apr 01 00:00:00 EDT 2013
Contribution of DOG1 expression to the diagnosis of gastrointestinal stromal tumors.
Pathology, research and practice
Kara T,Serinsoz E,Arpaci RB,Gubur O,Orekici G,Ata A,Colak T,Arican A
MA1-37028 was used in immunohistochemistry to study the diagnostic value of the immunohistochemical expression of DOG1 in gastrointestinal stromal tumors
Mon Jul 01 00:00:00 EDT 2013
Time-dependent expression and distribution of Egr-1 during skeletal muscle wound healing in rats.
Journal of molecular histology
Fan YY,Ye GH,Lin KZ,Yu LS,Wu SZ,Dong MW,Han JG,Feng XP,Li XB
MA1-37028 was used in immunohistochemistry to study the temporal and cellular expression of Egr-1 during wound repair in rat skeletal muscle
Fri Feb 01 00:00:00 EST 2013
Dural angioleiomyoma of the middle cranial fossa: a case report and review of the literature.
Brain tumor pathology
Zhou Z,Yu M,Yang S,Zhou J,Sun R,Yang G
MA1-37028 was used in immunohistochemistry to report on a case of dural angioleiomyoma of the middle cranial fossa
Mon Apr 01 00:00:00 EDT 2013
DNA microarray and quantitative analysis reveal enhanced myocardial VEGF expression with stunted angiogenesis in human tetralogy of Fallot.
Cell biochemistry and biophysics
Peters TH,Sharma V,Yilmaz E,Mooi WJ,Bogers AJ,Sharma HS
MA1-37028 was used in immunohistochemistry to study the role of elevated myocardial expression of VEGF in the pathology of human tetralogy of Fallot
Fri Nov 01 00:00:00 EDT 2013
The effects of hyperbaric oxygen application against cholestatic oxidative stress and hepatic damage after bile duct ligation in rats.
The Journal of surgical research
Ayvaz S,Kanter M,Aksu B,Sahin SH,Uzun H,Erboga M,Pul M
MA1-37028 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of hyperbaric oxygen against oxidative and cholestatic hepatic damage caused by bile duct ligation in rats
Mon Jul 01 00:00:00 EDT 2013
Stromal-epithelial crosstalk provides a suitable microenvironment for the progression of ovarian cancer cells in vitro.
Cancer investigation
Fu S,Dong L,Sun W,Xu Y,Gao L,Miao Y
MA1-37028 was used in immunocytochemistry to study the in vivo progression of ovarian cancer cells and the role of stromal-epithelial interactions in creating an appropriate microenvironment
Fri Nov 01 00:00:00 EDT 2013
Protective effect of Urtica dioica on liver damage induced by biliary obstruction in rats.
Toxicology and industrial health
Oguz S,Kanter M,Erboga M,Ibis C
MA1-37028 was used in immunohistochemistry to study the ability of Urtica dioica to protect against liver damage in a rat bile duct ligation model
Tue Oct 01 00:00:00 EDT 2013
Effect of mesenchymal stem cells on anti-Thy1,1 induced kidney injury in albino rats.
Asian Pacific journal of tropical biomedicine
Sakr S,Rashed L,Zarouk W,El-Shamy R
MA1-37028 was used in immunohistochemistry to study the beneficial effects in a rat model of nephritis of therapy with bone marrow-derived mesenchymal stem cells
Fri Mar 01 00:00:00 EST 2013
Mesenchymal-to-endothelial transition in Kaposi sarcoma: a histogenetic hypothesis based on a case series and literature review.
PloS one
Gurzu S,Ciortea D,Munteanu T,Kezdi-Zaharia I,Jung I
MA1-37028 was used in immunohistochemistry to develop a hypothesis of Kaposi sarcoma histogenesis based on a case and literature review
Tue Apr 01 00:00:00 EDT 2014
Infarct-induced steroidogenic acute regulatory protein: a survival role in cardiac fibroblasts.
Molecular endocrinology (Baltimore, Md.)
Anuka E,Yivgi-Ohana N,Eimerl S,Garfinkel B,Melamed-Book N,Chepurkol E,Aravot D,Zinman T,Shainberg A,Hochhauser E,Orly J
MA1-37028 was used in immunohistochemistry to study the role of StAR in protecting cardiac fibroblast from apoptosis in the post-infarct heart
Sun Sep 01 00:00:00 EDT 2013
Keratocyte apoptosis and not myofibroblast differentiation mark the graft/host interface at early time-points post-DSAEK in a cat model.
PloS one
Weis AJ,Huxlin KR,Callan CL,DeMagistris MA,Hindman HB
MA1-37028 was used in immunohistochemistry to study keratinocyte apoptosis as a graft-host marker in a feline model of Descemet's stripping automated endothelial keratoplasty
Mon May 12 00:00:00 EDT 2014
Isolation, culture and evaluation of multilineage-differentiating stress-enduring (Muse) cells.
Nature protocols
Kuroda Y,Wakao S,Kitada M,Murakami T,Nojima M,Dezawa M
MA1-37028 was used in immunocytochemistry to describe methods to isolate and evaluate multilineage-differentiating stress-enduring cells
Tue Jan 14 00:00:00 EST 2014
Diet induced mild hypercholesterolemia in pigs: local and systemic inflammation, effects on vascular injury - rescue by high-dose statin treatment.
PloS one
Busnelli M,Manzini S,Froio A,Vargiolu A,Cerrito MG,Smolenski RT,Giunti M,Cinti A,Zannoni A,Leone BE,Forni M,Bacci ML,Biasi GM,Giovannoni R,Lavitrano M
MA1-37028 was used in immunohistochemistry to study the effects of high-dose statin therapy on inflammation and vascular injury in a porcine model of mild hypercholesterolemia
Tue Aug 26 00:00:00 EDT 2014
Attenuation of CCl4-induced hepatic fibrosis in mice by vaccinating against TGF-β1.
PloS one
Fan X,Zhang Q,Li S,Lv Y,Su H,Jiang H,Hao Z
MA1-37028 was used in western blot to study the ability of vaccination against TGF-beta1 to protect against hepatic fibrosis in a liver injury model
Sun Oct 12 00:00:00 EDT 2014
Oral administration of recombinant adeno-associated virus-mediated bone morphogenetic protein-7 suppresses CCl(4)-induced hepatic fibrosis in mice.
Molecular therapy : the journal of the American Society of Gene Therapy
Hao ZM,Cai M,Lv YF,Huang YH,Li HH
MA1-37028 was used in immunohistochemistry to study the ability of an orally administered recombinant AAV-BMP-7 virus to protect against experimentally induced hepatic fibrosis in a murine model
Thu Nov 01 00:00:00 EDT 2012
Transgenic expression of interferon-γ in mouse stomach leads to inflammation, metaplasia, and dysplasia.
The American journal of pathology
Syu LJ,El-Zaatari M,Eaton KA,Liu Z,Tetarbe M,Keeley TM,Pero J,Ferris J,Wilbert D,Kaatz A,Zheng X,Qiao X,Grachtchouk M,Gumucio DL,Merchant JL,Samuelson LC,Dlugosz AA
MA1-37028 was used in immunohistochemistry to study the induction of inflammation, metaplasia and displasia following transgenic expression of IFN-gamma in the murine stomach
Sat Dec 01 00:00:00 EST 2012
Loss of semaphorin-neuropilin-1 signaling causes dysmorphic vascularization reminiscent of alveolar capillary dysplasia.
The American journal of pathology
Joza S,Wang J,Fox E,Hillman V,Ackerley C,Post M
MA1-37028 was used in immunohistochemistry to study the role of SEMA3-neuropilin-1 signaling in the vascular development of the fetal lung and the effects of signaling deficiency
Sat Dec 01 00:00:00 EST 2012
The pulmonary mesenchymal tissue layer is defective in an in vitro recombinant model of nitrofen-induced lung hypoplasia.
The American journal of pathology
van Loenhout RB,Tseu I,Fox EK,Huang Z,Tibboel D,Post M,Keijzer R
MA1-37028 was used in immunohistochemistry to develop a recombinant in vitro cellular model of lung hypoplasia
Sun Jan 01 00:00:00 EST 2012
Multipotent nestin-expressing stem cells capable of forming neurons are located in the upper, middle and lower part of the vibrissa hair follicle.
Cell cycle (Georgetown, Tex.)
Amoh Y,Mii S,Aki R,Hamada Y,Kawahara K,Hoffman RM,Katsuoka K
MA1-37028 was used in immunohistochemistry to study hair follicle multipotent stem cells that express nestin and are capable of differentiating into neurons
Sat Sep 15 00:00:00 EDT 2012
Asbestos exposure induces alveolar epithelial cell plasticity through MAPK/Erk signaling.
Journal of cellular biochemistry
Tamminen JA,Myllärniemi M,Hyytiäinen M,Keski-Oja J,Koli K
MA1-37028 was used in western blot to study the role of MAPK/Erk signaling in the mechanism by which asbestos exposure induces plasticity in alveolar epithelial cells
Sun Jul 01 00:00:00 EDT 2012
Vaccination with platelet-derived growth factor B kinoids inhibits CCl₄-induced hepatic fibrosis in mice.
The Journal of pharmacology and experimental therapeutics
Hao ZM,Fan XB,Li S,Lv YF,Su HQ,Jiang HP,Li HH
MA1-37028 was used in western blot to study the ability of vaccination against PDGF-D to protect against hepatic fibrosis in a rat model of acute liver injury
Sat Sep 01 00:00:00 EDT 2012
Fibroblast growth factor 8b causes progressive stromal and epithelial changes in the epididymis and degeneration of the seminiferous epithelium in the testis of transgenic mice.
Biology of reproduction
Elo T,Sipilä P,Valve E,Kujala P,Toppari J,Poutanen M,Härkönen P
MA1-37028 was used in immunohistochemistry to study the epididymal and testicular changes caused by overexpression of human FGF8a in transgenic mice
Tue May 01 00:00:00 EDT 2012
Regulation of TGF-β storage and activation in the human idiopathic pulmonary fibrosis lung.
Cell and tissue research
Leppäranta O,Sens C,Salmenkivi K,Kinnula VL,Keski-Oja J,Myllärniemi M,Koli K
MA1-37028 was used in immunohistochemistry to study the role of LTBP-1 in modulating TGF-beta levels in the lungs of patients with idiopathic pulmonary fibrosis
Fri Jun 01 00:00:00 EDT 2012
Protective effects of thymoquinone against cholestatic oxidative stress and hepatic damage after biliary obstruction in rats.
Journal of molecular histology
Oguz S,Kanter M,Erboga M,Erenoglu C
MA1-37028 was used in immunohistochemistry to study the mechanisms underlying the protective and therapeutic effects of thymoquinone in a rat model of cholestatic liver disease
Sun Apr 01 00:00:00 EDT 2012
Fstl1 antagonizes BMP signaling and regulates ureter development.
PloS one
Xu J,Qi X,Gong J,Yu M,Zhang F,Sha H,Gao X
MA1-37028 was used in immunocytochemistry to study the antagonization of BMP signaling by Fstl1 and the significance for normal development of the ureter
Fri Jul 27 00:00:00 EDT 2012
TDAG51 mediates epithelial-to-mesenchymal transition in human proximal tubular epithelium.
American journal of physiology. Renal physiology
Carlisle RE,Heffernan A,Brimble E,Liu L,Jerome D,Collins CA,Mohammed-Ali Z,Margetts PJ,Austin RC,Dickhout JG
MA1-37028 was used in immunocytochemistry to study the epithelial-mesenchymal transition in human proximal tubular epithelium and the role played by TDAG51
Wed Aug 01 00:00:00 EDT 2012
Accreta complicating complete placenta previa is characterized by reduced systemic levels of vascular endothelial growth factor and by epithelial-to-mesenchymal transition of the invasive trophoblast.
American journal of obstetrics and gynecology
Wehrum MJ,Buhimschi IA,Salafia C,Thung S,Bahtiyar MO,Werner EF,Campbell KH,Laky C,Sfakianaki AK,Zhao G,Funai EF,Buhimschi CS
MA1-37028 was used in immunohistochemistry to study the serum angiogenic factor profile of women with complete placenta previa
Sun May 01 00:00:00 EDT 2011
Influence of glycosaminoglycan identity on vocal fold fibroblast behavior.
Acta biomaterialia
Jimenez-Vergara AC,Munoz-Pinto DJ,Becerra-Bayona S,Wang B,Iacob A,Hahn MS
MA1-37028 was used in immunohistochemistry to study the role of glycosaminoglycan identity in vocal fold fibroblast behavior
Tue Nov 01 00:00:00 EDT 2011
Gastrointestinal stromal tumours in children and young adults: a clinicopathologic series with long-term follow-up from the database of the Cooperative Weichteilsarkom Studiengruppe (CWS).
European journal of cancer (Oxford, England : 1990)
Benesch M,Leuschner I,Wardelmann E,Thielen M,Schmid I,Kontny U,Ebetsberger G,Frey E,Graf N,Schneider DT,Kremens B,Amann G,Urban C,Schlemmer M,Quehenberger F,Klingebiel T,Dantonello T,Koscielniak E
MA1-37028 was used in immunohistochemistry to investigate gastrointestinal stromal tumours in patients with long-term follow-up
Fri Jul 01 00:00:00 EDT 2011
Carcinoma ex pleomorphic adenoma of the salivary glands: distinct clinicopathologic features and immunoprofiles between subgroups according to cellular differentiation.
Journal of Korean medical science
Kim JW,Kwon GY,Roh JL,Choi SH,Nam SY,Kim SY,Cho KJ
MA1-37028 was used in immunohistochemistry to study the clinicopathological implications and mechanisms of cellular differentiation in carcinoma ex pleomorphic adenoma
Sat Oct 01 00:00:00 EDT 2011
Mesenchymal stem cells improve the healing of ischemic colonic anastomoses (experimental study).
Langenbeck's archives of surgery
Adas G,Arikan S,Karatepe O,Kemik O,Ayhan S,Karaoz E,Kamali G,Eryasar B,Ustek D
MA1-37028 was used in immunohistochemistry to study the role of allogenic mesenchymal stem cell transplantation in improved healing of ischemic colon anastomosis
Sat Jan 01 00:00:00 EST 2011
Primary kaposiform hemangioendothelioma of a long bone: two cases in unusual locations with long-term follow up.
Pathology international
Ma J,Shi QL,Jiang SJ,Zhou HB,Zhou XJ
MA1-37028 was used in immunohistochemistry to report on two cases of primary kaposiform hemangioendothelioma of a long bone
Wed Jun 01 00:00:00 EDT 2011
Use of p63, a myoepithelial cell marker, in determining the invasiveness of spontaneous mammary neoplasia in a rhesus macaque (Macaca mulatta).
Journal of the American Association for Laboratory Animal Science : JAALAS
Williams-Fritze MJ,Carlson Scholz JA,Bossuyt V,Booth CJ
MA1-37028 was used in immunohistochemistry to investigate the effect of p63 on the invasiveness of breast cancer
Tue Mar 01 00:00:00 EST 2011
Papillary urothelial carcinoma of the bladder with exuberant pseudosarcomatous stromal reaction following radiation therapy.
International journal of surgical pathology
Ambrosini-Spaltro A,Melissari M
MA1-37028 was used in immunohistochemistry to report on a case of papillary urothelial carcinoma of the bladder with exuberant pseudosarcomatous stromal reaction
Fri Apr 01 00:00:00 EDT 2011
A case of prostatic adenocarcinoma with aberrant p63 expression: presentation with detailed immunohistochemical study and FISH analysis.
International journal of surgical pathology
Baydar DE,Kulac I,Gurel B,De Marzo A
MA1-37028 was used in immunohistochemistry to report a new clinical case of prostatic adenomas with abnormal p63 expression
Tue Feb 01 00:00:00 EST 2011
Histopathological and immunohistochemical features of gastrointestinal stromal tumors.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie
Fülöp E,Marcu S,Borda A,Moldovan C,Fülöp EF,Loghin A,Pávai Z
MA1-37028 was used in immunohistochemistry to characterize gastrointestinal stromal tumors
Wed Sep 28 00:00:00 EDT 2011
Primary malignant deciduoid peritoneal mesothelioma: a report of the cytohistological and immunohistochemical appearances.
Diagnostic cytopathology
Ustun H,Astarci HM,Sungu N,Ozdemir A,Ekinci C
MA1-37028 was used in immunohistochemistry to investigate deciduoid features of malignant mesothelioma by histomorphological and immunohistochemical evaluation
Wed Jun 01 00:00:00 EDT 2011
Cotyledonoid leiomyoma of uterus: a case report.
Turk patoloji dergisi
Ersöz S,Turgutalp H,Mungan S,Güvendı G,Güven S
MA1-37028 was used in immunohistochemistry to report on a case of cotyledonoid leiomyoma of uterus
Tue Jan 17 00:00:00 EST 2012
Fascin expression and its potential significance in gastrointestinal stromal tumors.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology
Ozcan A,Karslioğlu Y,Günal A,Cermık AH,Kurt B,Ongürü O
MA1-37028 was used in immunohistochemistry to study the expression of fascin in gastrointestinal stromal tumors and its possible prognostic value
Mon Aug 01 00:00:00 EDT 2011
Antagonistic roles of Notch and p63 in controlling mammary epithelial cell fates.
Cell death and differentiation
Yalcin-Ozuysal O,Fiche M,Guitierrez M,Wagner KU,Raffoul W,Brisken C
MA1-37028 was used in immunohistochemistry to investigate the role of Notch and p63 in regulation of mammary epithelial cell fates
Fri Oct 01 00:00:00 EDT 2010
Primary adenomyoepithelioma of the sellar region: a case report.
The American journal of surgical pathology
Rychly B,Kazakov DV,Danis D,Szep Z,Michal M
MA1-37028 was used in immunohistochemistry to characterize a specific case of sellar adenomyoepithelioma
Fri Oct 01 00:00:00 EDT 2010
Rescue treatment with a Rho-kinase inhibitor normalizes right ventricular function and reverses remodeling in juvenile rats with chronic pulmonary hypertension.
American journal of physiology. Heart and circulatory physiology
Xu EZ,Kantores C,Ivanovska J,Engelberts D,Kavanagh BP,McNamara PJ,Jankov RP
MA1-37028 was used in immunohistochemistry to investigate the effect of ROCK inhibition on chronic pulmonary hypertension in rats
Wed Dec 01 00:00:00 EST 2010
Spermatogonia-specific proteins expressed in prepubertal buffalo (Bubalus bubalis) testis and their utilization for isolation and in vitro cultivation of spermatogonia.
Theriogenology
Goel S,Reddy N,Mandal S,Fujihara M,Kim SM,Imai H
MA1-37028 was used in immunohistochemistry to investigate the culture system for buffalo spermatogonia
Fri Oct 15 00:00:00 EDT 2010
Assessment of the tumorigenesis and drug susceptibility of three new canine mammary tumor cell lines.
Research in veterinary science
Chang CY,Chiou PP,Chen WJ,Li YH,Yiu JC,Cheng YH,Chen SD,Lin CT,Lai YS
MA1-37028 was used in western blot to study the tumorigenesis and drug susceptibility of three new canine mammary tumor cell lines
Thu Apr 01 00:00:00 EDT 2010
Isolation and in vitro characterisation of dental pulp stem cells from natal teeth.
Histochemistry and cell biology
Karaöz E,Doğan BN,Aksoy A,Gacar G,Akyüz S,Ayhan S,Genç ZS,Yürüker S,Duruksu G,Demircan PC,Sariboyaci AE
MA1-37028 was used in immunocytochemistry to characterize the biological properties and differentiation potential of human natal dental pulp stem cells
Fri Jan 01 00:00:00 EST 2010
Pancreatic islet derived stem cells can express co-stimulatory molecules of antigen-presenting cells.
Transplantation proceedings
Karaoz E,Okçu A,Saglam O,Genc ZS,Ayhan S,Kasap M
MA1-37028 was used in immunohistochemistry to study the expression of antigen presenting cell co-stimulatory molecules by pancreatic islet derived stem cells
Mon Nov 01 00:00:00 EDT 2010
Immunohistochemical comparative study of fibrosis and biliary ductular reaction in alcoholic and viral chronic hepatitis.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie
Egyed-Zsigmond I,Jung I,Egyed-Zsigmond I,Marton G,Gurzu S,Mezei T
MA1-37028 was used in immunohistochemistry to investigate the characteristics of fibrosis in patients with alcoholic and viral chronic hepatitis
Tue Nov 30 00:00:00 EST 2010
Solitary fibrous tumor of soft tissue: a case report and immunohistochemical study.
Medical molecular morphology
Yuri T,Kanematsu S,Lei YC,Kuwata M,Oishi M,Tsubura A
MA1-37028 was used in immunohistochemistry to report a clinical case of solitary fibrous tumor of soft tissue
Mon Mar 01 00:00:00 EST 2010
Myosin light chain kinase is necessary for tonic airway smooth muscle contraction.
The Journal of biological chemistry
Zhang WC,Peng YJ,Zhang GS,He WQ,Qiao YN,Dong YY,Gao YQ,Chen C,Zhang CH,Li W,Shen HH,Ning W,Kamm KE,Stull JT,Gao X,Zhu MS
MA1-37028 was used in immunohistochemistry to study the essential role of myosin light chain kinase in tonic airway smooth muscle contraction
Fri Feb 19 00:00:00 EST 2010
Melanotic Xp11 translocation renal cancer: a case with PSF-TFE3 gene fusion and up-regulation of melanogenetic transcripts.
The American journal of surgical pathology
Chang IW,Huang HY,Sung MT
MA1-37028 was used in immunohistochemistry to report on a case of melanotic Xp11 translocation renal cancer
Tue Dec 01 00:00:00 EST 2009
Role of stem cell factor and bone marrow-derived fibroblasts in airway remodeling.
The American journal of pathology
Dolgachev VA,Ullenbruch MR,Lukacs NW,Phan SH
MA1-37028 was used in immunohistochemistry to investigate the effect of stem cell factor on airway remodeling and its mechanism
Sun Feb 01 00:00:00 EST 2009
Localization of mesenchymal cells in adult mouse thymus: their abnormal distribution in mice with disorganization of thymic medullary epithelium.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Odaka C
MA1-37028 was used in immunohistochemistry to examine the distribution of mesenchymal cells in normal and abnormal mouse thymus
Wed Apr 01 00:00:00 EDT 2009
Cyclooxygenase-2 expression and connection with tumor recurrence and histopathologic parameters in gastrointestinal stromal tumors.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Türköz HK,Alkan I,Sişman S,Ozcan D
MA1-37028 was used in immunohistochemistry to study the cyclooxygenase-2 expression and its effect on mesenchymal tumor growth and recurrence
Sun Nov 01 00:00:00 EDT 2009
Rhabdomyosarcoma arising in a mature cystic teratoma with contralateral serous carcinoma: case report and review of the literature.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Kefeli M,Kandemir B,Akpolat I,Yildirim A,Kokcu A
MA1-37028 was used in immunohistochemistry to report on a case of rhabdomyosarcoma arising in a mature cystic teratoma with contralateral serous carcinoma
Wed Jul 01 00:00:00 EDT 2009
Coexistence of different tissue tumourigenesis in an N-methyl-N-nitrosourea-induced mammary carcinoma model: a histopathological report in Sprague-Dawley rats.
Laboratory animals
Esendagli G,Yilmaz G,Canpinar H,Gunel-Ozcan A,Guc MO,Guc D
MA1-37028 was used in immunohistochemistry to investigate tumourigenesis in non-mammary tissues in the MNU-induced mammary tumour model
Thu Jan 01 00:00:00 EST 2009
Characterization of mesenchymal stem cells from rat bone marrow: ultrastructural properties, differentiation potential and immunophenotypic markers.
Histochemistry and cell biology
Karaoz E,Aksoy A,Ayhan S,Sariboyaci AE,Kaymaz F,Kasap M
MA1-37028 was used in immunocytochemistry to study the morphology and immunology of rat bone marrow-derived mesenchymal stem cells
Sun Nov 01 00:00:00 EDT 2009
Multipotential ability of primitive germ cells from neonatal pig testis cultured in vitro.
Reproduction, fertility, and development
Goel S,Fujihara M,Tsuchiya K,Takagi Y,Minami N,Yamada M,Imai H
MA1-37028 was used in immunocytochemistry to investigate the stem cell potential of primitive germ cells isolated from neonatal pig testis
Tue Aug 18 00:00:00 EDT 2009
Giant cell tumor-like lesion of the urinary bladder: a report of two cases and literature review; giant cell tumor or undifferentiated carcinoma?
Diagnostic pathology
Behzatoğlu K,Durak H,Canberk S,Aydin O,Huq GE,Oznur M,Ozyalvaçli G,Yildiz P
MA1-37028 was used in immunohistochemistry to report on two cases of giant cell tumor-like lesion of the urinary bladder
Thu Dec 31 00:00:00 EST 2009
Effect of visible and infrared polarized light on the healing process of full-thickness skin wounds: an experimental study.
Photomedicine and laser surgery
Iordanou P,Lykoudis EG,Athanasiou A,Koniaris E,Papaevangelou M,Fatsea T,Bellou P
MA1-37028 was used in immunohistochemistry to study the effect of visible and infrared polarized light on skin wound healing in rats
Wed Apr 01 00:00:00 EDT 2009
There are no estrogen and progesterone receptors in cerebral cavernomas: a preliminary immunohistochemical study.
Surgical neurology
Kaya AH,Ulus A,Bayri Y,Topal A,Gun S,Kandemir B,Dagcinar A,Senel A,Iyigun O
MA1-37028 was used in immunohistochemistry to study the absence of estrogen and progesterone receptors in cerebral cavernomas
Tue Sep 01 00:00:00 EDT 2009
Multiple primary sporadic gastrointestinal stromal tumors in the adult: an underestimated entity.
Clinical cancer research : an official journal of the American Association for Cancer Research
Gasparotto D,Rossi S,Bearzi I,Doglioni C,Marzotto A,Hornick JL,Grizzo A,Sartor C,Mandolesi A,Sciot R,Debiec-Rychter M,Dei Tos AP,Maestro R
MA1-37028 was used in immunohistochemistry to study the incidence of multiple primary sporadic gastrointestinal stromal tumors
Mon Sep 15 00:00:00 EDT 2008
Influence of hydrogel mechanical properties and mesh size on vocal fold fibroblast extracellular matrix production and phenotype.
Acta biomaterialia
Liao H,Munoz-Pinto D,Qu X,Hou Y,Grunlan MA,Hahn MS
MA1-37028 was used in immunohistochemistry to investigate the effect of PEGDA hydrogel on the production of extracellular matrix from vocal fold fibroblasts
Mon Sep 01 00:00:00 EDT 2008
Activation of estrogen signaling pathways collaborates with loss of Brca1 to promote development of ERalpha-negative and ERalpha-positive mammary preneoplasia and cancer.
Oncogene
Jones LP,Tilli MT,Assefnia S,Torre K,Halama ED,Parrish A,Rosen EM,Furth PA
MA1-37028 was used in immunohistochemistry - paraffin section to test if Brca1 loss in mammary epithelium alters the estrogenic growth response and if exposure to increased estrogen or ERalpha correlates with Brca1 deficiency
Thu Jan 31 00:00:00 EST 2008
Chick pulmonary Wnt5a directs airway and vascular tubulogenesis.
Development (Cambridge, England)
Loscertales M,Mikels AJ,Hu JK,Donahoe PK,Roberts DJ
MA1-37028 was used in immunohistochemistry to study the role of non-cannonical Wnt5a in modulating pulmonary airway and vascular tubulogenesis
Tue Apr 01 00:00:00 EDT 2008
Sarcoma of follicular dendritic cells with features of sinus lining cells--a new subtype of reticulum cell sarcoma?
Virchows Archiv : an international journal of pathology
Krokowski M,Merz H,Thorns C,Bernd HW,Schade U,Le Tourneau A,Diebold J,Feller AC
MA1-37028 was used in immunohistochemistry to characterize a potentially new subtype of reticulum cell sarcoma
Thu May 01 00:00:00 EDT 2008
Bone marrow-derived cells participate in stromal remodeling of the lung following acute bacterial pneumonia in mice.
Lung
Serikov VB,Mikhaylov VM,Krasnodembskay AD,Matthay MA
MA1-37028 was used in immunohistochemistry to study the role of bone marrow-derived cells in stromal remodeling of the lung following acute bacterial pneumonia in mice
Tue Nov 04 00:00:00 EST 2008
Vascular morphogenesis by adult bone marrow progenitor cells in three-dimensional fibrin matrices.
Differentiation; research in biological diversity
Rüger BM,Breuss J,Hollemann D,Yanagida G,Fischer MB,Mosberger I,Chott A,Lang I,Davis PF,Höcker P,Dettke M
MA1-37028 was used in immunohistochemistry to study the ability of human bone marrow progenitor cells to create vascular structures within 3D fibrin matrices
Mon Sep 01 00:00:00 EDT 2008
Chronic high-fat diets induce oxide injuries and fibrogenesis of pancreatic cells in rats.
Pancreas
Zhang X,Cui Y,Fang L,Li F
MA1-37028 was used in immunohistochemistry to investigate the effect of chronic high-fat diets on increasing pancreatic free fatty acids and lipid peroxidation
Wed Oct 01 00:00:00 EDT 2008
Culture of porcine fetal pancreatic neurons.
Pancreas
Kuroki M,Sawasaki Y,Kuroki K,Takeuchi K,Yoshioka M
MA1-37028 was used in immunocytochemistry to explore the culture of pancreatic neurons from porcine fetus
Fri Aug 01 00:00:00 EDT 2008
Indole derivatives sustain embryonic stem cell self-renewal in long-term culture.
Bioscience, biotechnology, and biochemistry
Miyabayashi T,Yamamoto M,Sato A,Sakano S,Takahashi Y
MA1-37028 was used in immunocytochemistry to study the role of indole derivatives in sustaining embryonic stem cell self-renewal in long-term culture
Thu May 01 00:00:00 EDT 2008
Glomeruloid hemangioma.
Pathology international
Yuri T,Yamazaki F,Takasu K,Shikata N,Tsubura A
MA1-37028 was used in immunohistochemistry to report on a case of glomeruloid hemangioma
Sun Jun 01 00:00:00 EDT 2008
Hepatic stellate cell activation and hepatic fibrosis induced by ischemia/reperfusion injury.
Transplantation proceedings
Cheng F,Li Y,Feng L,Li S
MA1-37028 was used in immunohistochemistry to study hepatic stellate cell activation and hepatic fibrosis induced by ischemia/reperfusion injury
Mon Sep 01 00:00:00 EDT 2008
Dedifferentiated Liposarcoma of the Retroperitoneum with Extensive Leiomyosarcomatous Differentiation and beta-Human Chorionic Gonadotropin Production.
Sarcoma
Russell MJ,Flynt FL,Harroff AL,Fadare O
MA1-37028 was used in immunohistochemistry to report a clinical case of dedifferentiated liposarcoma of the retroperitoneum with unique features
Thu Jul 14 00:00:00 EDT 2011
A stable explant culture of HER2/neu invasive carcinoma supported by alpha-Smooth Muscle Actin expressing stromal cells to evaluate therapeutic agents.
BMC cancer
Piechocki MP
MA1-37028 was used in flow cytometry to develop a co-culture model from an invasive lobular carcinoma for the evaluation of therapeutic agents
Thu Apr 24 00:00:00 EDT 2008
Treatment with imatinib improves drug delivery and efficacy in NSCLC xenografts.
British journal of cancer
Vlahovic G,Ponce AM,Rabbani Z,Salahuddin FK,Zgonjanin L,Spasojevic I,Vujaskovic Z,Dewhirst MW
MA1-37028 was used in immunohistochemistry to investigate the effect of imatinib treatment on drug delivery and efficacy in non-small cancer lung cell xenografts
Mon Sep 17 00:00:00 EDT 2007
The effects of cyclical oestrogen on bladder and urethral structure and function.
BJU international
Palmieri K,Mannikarottu AS,Chichester P,Kogan B,Leggett RE,Whitbeck C,Levin RM
MA1-37028 was used in immunohistochemistry to study the effects of cyclical oestrogen on bladder and urethral structure and function
Mon Jan 01 00:00:00 EST 2007
Identification, isolation, and in vitro culture of porcine gonocytes.
Biology of reproduction
Goel S,Sugimoto M,Minami N,Yamada M,Kume S,Imai H
MA1-37028 was used in immunohistochemistry to identify and characterize porcine gonocytes and establish in vitro culture
Sun Jul 01 00:00:00 EDT 2007
Elastofibroma: a clinicopathologic and immunohistochemical study of seven cases and literature review.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Gun BD,Bahadir B,Behzatoglu K,Gun MO,Ozdamar SO
MA1-37028 was used in immunohistochemistry to report on seven cases of elastofibroma
Thu Feb 01 00:00:00 EST 2007
Allopurinol in rat chronic pancreatitis: effects on pancreatic stellate cell activation.
Pancreas
Tasci I,Deveci S,Isik AT,Comert B,Akay C,Mas N,Inal V,Yamanel L,Mas MR
MA1-37028 was used in immunohistochemistry to investigate the effect of allopurinol on pancreatic stellate cells and its application for pancreatitis treatment
Thu Nov 01 00:00:00 EDT 2007
Increased expression of HIF-1alpha, nNOS, and VEGF in the cerebral cortex of anemic rats.
American journal of physiology. Regulatory, integrative and comparative physiology
McLaren AT,Marsden PA,Mazer CD,Baker AJ,Stewart DJ,Tsui AK,Li X,Yucel Y,Robb M,Boyd SR,Liu E,Yu J,Hare GM
MA1-37028 was used in immunocytochemistry to examine the effect of acute anemia on the expression of HIF-1alpha, nNOS, and VEGF in the mouse brain
Mon Jan 01 00:00:00 EST 2007
Epithelioid leiomyosarcoma with rhabdoid features.
Wiener klinische Wochenschrift
Yorulmaz G,Erdogan G,Pestereli HE,Savas B,Karaveli FS
MA1-37028 was used in immunohistochemistry to report on a case of epithelioid leiomyosarcoma with rhabdoid features
Wed Jan 30 00:00:00 EST 2008
Evidence of temporary airway epithelial repopulation and rare clonal formation by BM-derived cells following naphthalene injury in mice.
Anatomical record (Hoboken, N.J. : 2007)
Serikov VB,Popov B,Mikhailov VM,Gupta N,Matthay MA
MA1-37028 was used in immunohistochemistry to study the involvement of bone marrow cells in airway epithelial restoration following naphthalene-induced injury
Sat Sep 01 00:00:00 EDT 2007
Myointimoma of the glans penis.
Pathology international
Vardar E,Gunlusoy B,Arslan M,Kececi S
MA1-37028 was used in immunohistochemistry to report on a case of myointimoma of the glans penis
Thu Mar 01 00:00:00 EST 2007
Vulvar leiomyosarcoma in a cat.
Journal of feline medicine and surgery
Firat I,Haktanir-Yatkin D,Sontas BH,Ekici H
MA1-37028 was used in immunohistochemistry to report on a case of vulvar leiomyosarcoma in a cat
Mon Oct 01 00:00:00 EDT 2007
Ex vivo reversal of in vivo transdifferentiation in mesothelial cells grown from peritoneal dialysate effluents.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
Vargha R,Endemann M,Kratochwill K,Riesenhuber A,Wick N,Krachler AM,Malaga-Dieguez L,Aufricht C
MA1-37028 was used in western blot to study the ex vivo reversal of in vivo transdifferentiation of mesothelial cells
Sun Oct 01 00:00:00 EDT 2006
PAR3 is essential for cyst-mediated epicardial development by establishing apical cortical domains.
Development (Cambridge, England)
Hirose T,Karasawa M,Sugitani Y,Fujisawa M,Akimoto K,Ohno S,Noda T
MA1-37028 was used in immunohistochemistry - paraffin section to report that disruption of the mouse Par3 gene results in midgestational embryonic lethality with defective epicardial development
Sat Apr 01 00:00:00 EST 2006
Microkeratome assisted deep lamellar keratoprosthesis.
The British journal of ophthalmology
Shimmura S,Miyashita H,Uchino Y,Taguchi T,Kobayashi H,Shimazaki J,Tanaka J,Tsubota K
MA1-37028 was used in immunohistochemistry to evaluate a new surgical cornea replacement procedure
Sat Jul 01 00:00:00 EDT 2006
Biological coating for arterial stents: the next evolutionary change in stents.
Journal of endovascular therapy : an official journal of the International Society of Endovascular Specialists
Carnevale K,Ouriel K,Gabriel Y,Clair D,Bena JF,Silva MB,Sarac TP
MA1-37028 was used in immunohistochemistry to study the use of peritoneum in stent linings
Sat Apr 01 00:00:00 EST 2006
Long-term effect of experimental hypercholesterolemia on cavernosal tissues.
Urology
Karaboga R,Kilic O,Yaman O,Percinel S,Anafarta K
MA1-37028 was used in immunohistochemistry to determine the effect of long-term experimental hypercholesterolemia on cavernosal tissues
Wed Feb 01 00:00:00 EST 2006
Immunohistochemical study of NG2 chondroitin sulfate proteoglycan expression in the small and large intestines.
Histochemistry and cell biology
Terada N,Ohno N,Murata S,Katoh R,Stallcup WB,Ohno S
MA1-37028 was used in immunohistochemistry to study NG2 proteoglycan expresssion in the small and large intestine
Sun Oct 01 00:00:00 EDT 2006
An in vitro model of trophoblast invasion of spiral arteries.
Methods in molecular medicine
Cartwright JE,Wareing M
MA1-37028 was used in immunohistochemistry to develop an in vitro model of trophoblast invasion of spiral arteries
Tue Apr 25 00:00:00 EDT 2006
The role of CXCL12 in the organ-specific process of artery formation.
Blood
Ara T,Tokoyoda K,Okamoto R,Koni PA,Nagasawa T
MA1-37028 was used in immunohistochemistry to study the role of CXCL12 in organ vascularization
Fri Apr 15 00:00:00 EDT 2005
Small bowel cyclooxygenase 2 (COX-2) expression in patients with IgA nephropathy.
Kidney international
Honkanen T,Mustonen J,Kainulainen H,Myllymäki J,Collin P,Hurme M,Rantala I
MA1-37028 was used in immunohistochemistry to investigate the association between small bowel COX-2 expression and IgA nephropathy
Wed Jun 01 00:00:00 EDT 2005
Renoprotective properties of angiotensin receptor blockers beyond blood pressure lowering.
Journal of the American Society of Nephrology : JASN
Izuhara Y,Nangaku M,Inagi R,Tominaga N,Aizawa T,Kurokawa K,van Ypersele de Strihou C,Miyata T
MA1-37028 was used in immunohistochemistry to study the ability of angiotensin receptor blockers to protect against kidney damage independently of blood pressure lowering effects
Thu Dec 01 00:00:00 EST 2005
Structural changes to airway smooth muscle in cystic fibrosis.
Thorax
Hays SR,Ferrando RE,Carter R,Wong HH,Woodruff PG
MA1-37028 was used in immunohistochemistry to study the structural changes to airway smooth muscle in cystic fibrosis
Tue Mar 01 00:00:00 EST 2005
Activation of the neuregulin-1/ErbB signaling pathway promotes the proliferation of neoplastic Schwann cells in human malignant peripheral nerve sheath tumors.
Oncogene
Stonecypher MS,Byer SJ,Grizzle WE,Carroll SL
MA1-37028 was used in immunocytochemistry to study the role of neuregulin-1/ErbB pathway activation in neoplastic Schwann cell proliferation in peripheral nerve sheath tumors.
Thu Aug 25 00:00:00 EDT 2005
Reduction of postischemic immune inflammatory response: an effective strategy for attenuating chronic allograft nephropathy.
Transplantation
Herrero-Fresneda I,Torras J,Vidal A,Lloberas N,Cruzado JM,Grinyó JM
MA1-37028 was used in immunohistochemistry to study reduction of postischemic immune inflammatory responses as a way of reducing chronic allograft nephropathy
Thu Jan 27 00:00:00 EST 2005
Analysis of prognostic and immunohistochemical factors in gastrointestinal stromal tumors with malignant potential.
Journal of gastrointestinal surgery : official journal of the Society for Surgery of the Alimentary Tract
Ozgüç H,Yilmazlar T,Yerci O,Soylu R,Tümay V,Filiz G,Zorluoglu A
MA1-37028 was used in immunohistochemistry to evaluate the prognostic significance of multiple factors in gastrointestinal stromal tumors with malignant potential
Tue Mar 01 00:00:00 EST 2005
Cystic lymphangioma of the right adrenal gland.
Pathology oncology research : POR
Ates LE,Kapran Y,Erbil Y,Barbaros U,Dizdaroglu F
MA1-37028 was used in immunohistochemistry to report a clinical case of cystic lymphangioma in the right adrenal gland
Tue Mar 21 00:00:00 EST 2006
Mesenchymal hamartoma of the liver in adulthood: immunohistochemical profiles, clinical and histopathological features in two patients.
Journal of hepato-biliary-pancreatic surgery
Yesim G,Gupse T,Zafer U,Ahmet A
MA1-37028 was used in immunohistochemistry to study two adult patients with mesenchymal hamartoma of the liver
Thu Jul 06 00:00:00 EDT 2006
Hyperplasia of smooth muscle in mild to moderate asthma without changes in cell size or gene expression.
American journal of respiratory and critical care medicine
Woodruff PG,Dolganov GM,Ferrando RE,Donnelly S,Hays SR,Solberg OD,Carter R,Wong HH,Cadbury PS,Fahy JV
MA1-37028 was used in immunohistochemistry to study smooth muscle changes in mild to moderate asthma
Sat May 01 00:00:00 EDT 2004
Bone marrow-derived progenitor cells in pulmonary fibrosis.
The Journal of clinical investigation
Hashimoto N,Jin H,Liu T,Chensue SW,Phan SH
MA1-37028 was used in immunocytochemistry to investigate the origine and role of bone marrow-derived progenitor cells in pulmonary fibrosis
Thu Jan 01 00:00:00 EST 2004
Methodologic advancements in the study of airway smooth muscle.
The Journal of allergy and clinical immunology
Kotlikoff MI,Kannan MS,Solway J,Deng KY,Deshpande DA,Dowell M,Feldman M,Green KS,Ji G,Johnston R,Lakser O,Lee J,Lund FE,Milla C,Mitchell RW,Nakai J,Rishniw M,Walseth TF,White TA,Wilson J,Xin HB,Woodruff PG
MA1-37028 was used in immunohistochemistry to evaluate and discuss the research methods for the study of airway smooth muscle
Sun Aug 01 00:00:00 EDT 2004
Glomerular structural and functional changes in a high-fat diet mouse model of early-stage Type 2 diabetes.
Diabetologia
Wei P,Lane PH,Lane JT,Padanilam BJ,Sansom SC
MA1-37028 was used in immunohistochemistry to study glomerular structural and functional changes in a high-fat diet mouse model of early-stage Type 2 diabetes
Wed Sep 01 00:00:00 EDT 2004
Smad3 signaling is required for epithelial-mesenchymal transition of lens epithelium after injury.
The American journal of pathology
Saika S,Kono-Saika S,Ohnishi Y,Sato M,Muragaki Y,Ooshima A,Flanders KC,Yoo J,Anzano M,Liu CY,Kao WW,Roberts AB
MA1-37028 was used in western blot to study the role of msad3 signaling in epithelial-mesenchymal transition of lens epithelium during wound healing
Sun Feb 01 00:00:00 EST 2004
Ginkgo biloba extract reverses CCl4-induced liver fibrosis in rats.
World journal of gastroenterology
Luo YJ,Yu JP,Shi ZH,Wang L
MA1-37028 was used in immunohistochemistry to study the potential mechanisms by which gonko biloba extract reverses CCl4-induced liver injury in a mouse model
Thu Apr 01 00:00:00 EST 2004
Malignant gastrointestinal stromal tumor of the gallbladder.
Journal of Korean medical science
Park JK,Choi SH,Lee S,Min KO,Yun SS,Jeon HM
MA1-37028 was used in immunohistochemistry to report on a case of malignant gastrointestinal stromal tumor of the gallbladder
Fri Oct 01 00:00:00 EDT 2004
Mammary ECM composition and function are altered by reproductive state.
Molecular carcinogenesis
Schedin P,Mitrenga T,McDaniel S,Kaeck M
MA1-37028 was used in immunohistochemistry to study the effect of reproductive state on mammary gland extracellular matrix composition and function
Wed Dec 01 00:00:00 EST 2004
Collagenous fibroma (desmoplastic fibroblastoma).
Yonsei medical journal
Dagli M,Eryilmaz A,Acar A,Kulacoglu S,Akmansu H
MA1-37028 was used in immunohistochemistry to report on a patient with collagenous fibroma
Sun Oct 31 00:00:00 EDT 2004
Synchronous primary adenocarcinoma and gastrointestinal stromal tumor in the stomach: a report of two cases.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology
Bircan S,Candir O,Aydin S,Başpinar S,Bülbül M,Kapucuoğlu N,Karahan N,Ciriş M
MA1-37028 was used in immunohistochemistry to report on two patients with synchronous primary adenocarcinoma and gastrointestinal stromal tumor in the stomach
Wed Sep 01 00:00:00 EDT 2004
Effective contribution of transplanted vascular progenitor cells derived from embryonic stem cells to adult neovascularization in proper differentiation stage.
Blood
Yurugi-Kobayashi T,Itoh H,Yamashita J,Yamahara K,Hirai H,Kobayashi T,Ogawa M,Nishikawa S,Nishikawa S,Nakao K
MA1-37028 was used in flow cytometry to study the contribution of transplanted of embryonic stem cell-derived vascular progenitor cells to adult neovascularization
Tue Apr 01 00:00:00 EST 2003
ALK-ATIC fusion in urinary bladder inflammatory myofibroblastic tumor.
Genes, chromosomes & cancer
Debiec-Rychter M,Marynen P,Hagemeijer A,Pauwels P
MA1-37028 was used in immunohistochemistry to report molecular and clinicopathologic features of urinary bladder inflammatory myofibroblastic tumor
Wed Oct 01 00:00:00 EDT 2003
TGF-beta1 induces aberrant laminin chain and collagen type IV isotype expression in the glomerular basement membrane.
Nephron. Experimental nephrology
Chai Q,Krag S,Miner JH,Nyengaard JR,Chai S,Wogensen L
MA1-37028 was used in immunohistochemistry to study the effect of TGF-beta1 on laminin and collagen expression in glomerular basement membrane
Tue Apr 04 00:00:00 EDT 2006
Apc modulates embryonic stem-cell differentiation by controlling the dosage of beta-catenin signaling.
Nature genetics
Kielman MF,Rindapää M,Gaspar C,van Poppel N,Breukel C,van Leeuwen S,Taketo MM,Roberts S,Smits R,Fodde R
MA1-37028 was used in immunohistochemistry to study the mechanism by which the APC modulates embryonic stem-cell differentiation
Sun Dec 01 00:00:00 EST 2002
Enterovirus replication in valvular tissue from patients with chronic rheumatic heart disease.
European heart journal
Li Y,Pan Z,Ji Y,Peng T,Archard LC,Zhang H
MA1-37028 was used in immunohistochemistry to study Enterovirus replication in valvular tissue from patients with chronic rheumatic heart disease
Mon Apr 01 00:00:00 EST 2002
Enhanced expression of fibroblast growth factors and receptor FGFR-1 during vascular remodeling in chronic obstructive pulmonary disease.
American journal of respiratory cell and molecular biology
Kranenburg AR,De Boer WI,Van Krieken JH,Mooi WJ,Walters JE,Saxena PR,Sterk PJ,Sharma HS
MA1-37028 was used in immunohistochemistry to study the role of fibroblast growth factors and FGFR-1 during vascular remodeling in COPD
Fri Nov 01 00:00:00 EST 2002
Mesangial expansion associated with glomerular endothelial cell activation and macrophage recruitment is developing in hyperlipidaemic apoE null mice.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
Bruneval P,Bariéty J,Bélair MF,Mandet C,Heudes D,Nicoletti A
MA1-37028 was used in immunohistochemistry to study the role of severe hypercholesterolaemia in the pathology of glomerula injury
Sun Dec 01 00:00:00 EST 2002
Two intraligamentary lipomatous tumors with immunohistochemical features.
Acta obstetricia et gynecologica Scandinavica
Cinel L,Düşmez D,Nabaei SH,Taner D,Pata O
MA1-37028 was used in immunohistochemistry to report on two cases of two intraligamentary lipomatous tumors
Thu Aug 01 00:00:00 EDT 2002
Tissue response to N-butyl-2-cyanoacrylate after percutaneous injection into cutaneous vascular lesions.
Annals of plastic surgery
Canter HI,Vargel I,Mavlll ME,Gököz A,Erk Y
MA1-37028 was used in immunohistochemistry to study tissue responses to percutaneous injection of an adhesive widely used in surgical procedures
Fri Nov 01 00:00:00 EST 2002
Inflammation is probably not a prerequisite for renal interstitial fibrosis in normoglycemic obese rats.
American journal of physiology. Renal physiology
Lavaud S,Poirier B,Mandet C,Bélair MF,Irinopoulou T,Heudes D,Bazin R,Bariéty J,Myara I,Chevalier J
MA1-37028 was used in immunohistochemistry to study the role of inflammation in the development of renal interstitial fibrosis in normoglycemic Zucker obese rats
Sun Apr 01 00:00:00 EST 2001