Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-10868 detects mouse Sperm Protein sp56 from mouse samples. This antibody does not cross react with human sperm.
MA1-10868 has been successfully used in ELISA, Western blot, and immunohistochemistry applications. This antibody will detect denatured, non-reduced sp56, but will not detect the denatured, reduced protein.
The MA1-10868 immunogen is a mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC122773.1; sperm fertilization protein 56; zona pellucida sperm-binding protein 3 receptor
Gene Aliases: AV264399; SP56
Entrez Gene ID: (Mouse) 22789
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support