Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
MA1-10866 detects mouse sperm protein sp56 from mouse and hamster samples. MA1-10866 does not cross-react with human sperm.
MA1-10866 has been successfully used in ELISA, Western blot, and immunohistochemistry applications. MA1-18066 will react with denatured, non-reduced sp56 but not with denatured, reduced sp56.
The MA1-10866 immunogen is a mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC122773.1; sperm fertilization protein 56; zona pellucida sperm-binding protein 3 receptor
Gene Aliases: AV264399; SP56
Entrez Gene ID: (Mouse) 22789
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support